Skip to Content
Merck
All Photos(2)

Key Documents

D115207

Sigma-Aldrich

6-Methyluracil

97%

Synonym(s):

2,4-Dihydroxy-6-methylpyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
Beilstein:
115647
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

318 °C (dec.) (lit.)

SMILES string

CC1=CC(=O)NC(=O)N1

InChI

1S/C5H6N2O2/c1-3-2-4(8)7-5(9)6-3/h2H,1H3,(H2,6,7,8,9)

InChI key

SHVCSCWHWMSGTE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of tetraalkylammonium derivatives of 6-methyluracil on the endplate potentials of muscles of different functional types.
I V Kovyazina et al.
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections, 399, 458-460 (2005-02-19)
Adrian Häberli et al.
Nucleosides, nucleotides & nucleic acids, 22(5-8), 1187-1189 (2003-10-21)
We synthesized pyrrolidino-C-nucleosides, incorporated them into oligodeoxynucleotides and investigated their pairing properties. The thermal duplex and triplex stabilities were measured. While triplex formation is destabilized in the case of pyrrolidino-pseudo-U and -T, pyrrolidino-pseudo-iso-C leads to an increase of the Tm
H Eshtiagh-Hosseini et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 78(5), 1392-1396 (2011-02-18)
This paper reports the hydrothermal synthesis, experimental and theoretical studies of a novel cocrystal compound in the 2:1 stoichiometric ratio of 6-methyluracil (6mu) and dipicolinic acid (pydcH(2)) formulated as [6mu](2)[pydcH(2)] (1), for the first time. DFT calculations were performed to
Lars Holland et al.
Inorganic chemistry, 46(26), 11356-11365 (2007-12-01)
The reaction of K2PtCl4 with an excess of 1-methyluracilate (1-MeU) in water at 60 degrees C leads to the formation of two major products, K2[Pt(1-MeU-N3)4].10H2O (1) and trans-K[Pt(1-MeU-N3)2(1-MeU-C5)(H2O)].3H2O (2). Addition of CuCl2 to an aqueous solution of 2 yields the
Zhiqiang Guo et al.
Bioorganic & medicinal chemistry letters, 13(19), 3311-3315 (2003-09-03)
The novel synthesis and SAR studies of 6-methyluracils as human GnRH receptor antagonists are discussed. Introduction of a small methyl substituent at the beta-position from N3 of the uracil improved the GnRH binding potency by 5- to 10-fold. The best

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service