Skip to Content
Merck
All Photos(1)

Key Documents

T0376

Sigma-Aldrich

Thymine

≥99%

Synonym(s):

2,4-Dihydroxy-5-methylpyrimidine, 5-Methyluracil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
Beilstein:
117880
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

Assay

≥99%

form

powder

mp

~320 °C (dec.) (lit.)

SMILES string

CC1=CNC(=O)NC1=O

InChI

1S/C5H6N2O2/c1-3-2-6-5(9)7-4(3)8/h2H,1H3,(H2,6,7,8,9)

InChI key

RWQNBRDOKXIBIV-UHFFFAOYSA-N

Gene Information

mouse ... Tymp(72962)

Looking for similar products? Visit Product Comparison Guide

General description

Thymine is one of the four nucleobases, along with adenine, guanine and cytosine found in deoxyribonucleic acids (DNA).

Application

Thymine has been used as a standard nitrogenous base in high-performance liquid chromatography-ultraviolet (HPLC-UV) for the quantification of bone DNA samples, Raman scattering experiments. It has also been used as supplement in C2C12 myoblast cells. Thymine may be used to study chemical processes that affect DNA structure, such a radiation induced radical production leading to base cross-linking reactions and derivitizations. It may be used to study the parameters of hydrogen bonding kinetics and energies with other nucleobases such as adenine.Thymine is used to develop sensitive heavy metal (mercury) detectors based on coordination chemistry and nanoparticle structures.

Biochem/physiol Actions

Thymine used in studying DNA structure byirradtaion methods leading to cross-linking reactions and derivitizations. Thymine dimers are indicative of DNA damage. Thymine is used with metal (mercury) to form thymine−HgII−thymine (T−HgII−T) duplexes. Thymine starvation in bacteria leads to halt in DNA synthesis and is referred as thymine-less death.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methods for detection of cytosine and thymine modifications in DNA
Berney M and McGouran JF
Nature Reviews Chemistry, 1-1 (2018)
Andrej Dementjev et al.
Scientific reports, 10(1), 17097-17097 (2020-10-15)
Identification of chemically homologous microcrystals in a polycrystal sample is a big challenge and requires developing specific highly sensitive tools. Second harmonic (SHG) and coherent anti-Stokes Raman scattering (CARS) spectroscopy can be used to reveal arrangement of thymine molecules, one
Yang-Wei Lin et al.
The Analyst, 136(5), 863-871 (2010-12-16)
Monitoring the levels of potentially toxic metal (PTM) ions (e.g., Hg(2+), Pb(2+), Cu(2+)) in aquatic ecosystems is important because these ions can have severe effects on human health and the environment. Gold (Au) nanomaterials are attractive sensing materials because of
MercuryII-mediated formation of thymine- HgII- thymine base pairs in DNA duplexes
Miyake Y, et al.
Journal of the American Chemical Society, 128(7), 2172-2173 (2006)
Quantification of nitrogenous bases, DNA and Collagen type I for the estimation of the postmortem interval in bone remains
Perez-Martinez C, et al.
Forensic Science International, 281, 106-112 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service