Skip to Content
Merck
All Photos(1)

Key Documents

CDS003382

Sigma-Aldrich

Tri-m-tolyl phosphate

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21O4P
CAS Number:
Molecular Weight:
368.36
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

Cc1cccc(OP(=O)(Oc2cccc(C)c2)Oc3cccc(C)c3)c1

InChI

1S/C21H21O4P/c1-16-7-4-10-19(13-16)23-26(22,24-20-11-5-8-17(2)14-20)25-21-12-6-9-18(3)15-21/h4-15H,1-3H3

InChI key

RMLPZKRPSQVRAB-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel J Duarte et al.
Neurotoxicology, 59, 222-230 (2016-02-07)
Exposure to tricresyl phosphates (TCPs), via for example contaminated cabin air, has been associated with health effects including the so-called aerotoxic syndrome. While TCP neurotoxicity is mainly attributed to ortho-isomers like tri-ortho-cresyl phosphate (ToCP), recent exposure and risk assessments indicate
Xiang Ge et al.
Environment international, 139, 105741-105741 (2020-04-20)
Electronic waste (e-waste) dismantling is an important source of flame retardant emissions, and may have potentially adverse effects on surrounding area. This study investigated their influence on the surrounding area and the human health risks after an industrial park was
Bo Peng et al.
Environment international, 135, 105348-105348 (2019-12-31)
A total of 41 play mats made from different raw materials, such as polyethylene (PE), ethylene-vinyl acetate copolymer (EVA), chemical crosslinked polyethylene (XPE), and polyvinyl chloride (PVC), were obtained from Chinese markets and analyzed for flame retardants. Polybrominated diphenyl ethers
Long Pang et al.
Environmental science and pollution research international, 26(27), 27862-27871 (2019-07-26)
In this study, nine OPEs were measured in 60 street dust samples collected from the central province of Henan, China. The total concentrations of OPEs (∑9OPEs) in street dust range from 2.77 to 505 μg/kg (median 59.3 μg/kg). Chlorinated OPEs are dominant
Marina G Pintado-Herrera et al.
Journal of chromatography. A, 1429, 107-118 (2016-01-10)
The main goal of this work was to develop, optimize and validate a multi-residue method for the simultaneous determination of 97 contaminants, including fragrances, UV filters, repellents, endocrine disruptors, biocides, polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), organophosphorus flame retardants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service