Skip to Content
Merck
All Photos(1)

Key Documents

CDS000449

Sigma-Aldrich

N,N-Diallylamine hydrochloride

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12ClN
Molecular Weight:
133.62
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

form

solid

SMILES string

Cl.C=CCNCC=C

InChI

1S/C6H11N.ClH/c1-3-5-7-6-4-2;/h3-4,7H,1-2,5-6H2;1H

InChI key

PZNOBXVHZYGUEX-UHFFFAOYSA-N

Related Categories

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shujuan Hu et al.
Materials (Basel, Switzerland), 12(6) (2019-03-21)
An innovative approach for fabricating porous alumina ceramics is demonstrated in this paper. The distinguished feature is that the construction of the porous structure stems from the interaction between ceramic particles, which is a poorly explored area. By tailoring the
Yang Peng et al.
Bioresource technology, 243, 628-633 (2017-07-15)
Considering the high concentration of mercury in industrial wastewater, such as coal-fired power plants and gold mining wastewater, this research study investigated the coupling process of lipid production and mercury bioremediation using microalgae cells. Chlorella vulgaris modified by biomimetic mineralization.
Jingjing Li et al.
International journal of nanomedicine, 13, 4607-4625 (2018-08-22)
The development of T1-T2 dual contrast agent (CA) favors the visualization of the lesion in a more accurate and reliable manner by magnetic resonance imaging (MRI). The relaxivity and the interference between T1 and T2 CA are the main concerns
Diego Gomez-Maldonado et al.
Polymers, 11(12) (2019-12-18)
With increasing global water temperatures and nutrient runoff in recent decades, the blooming season of algae lasts longer, resulting in toxin concentrations that exceed safe limits for human consumption and for recreational use. From the different toxins, microcystin-LR has been
Pedro V A Bueno et al.
International journal of biological macromolecules, 115, 792-800 (2018-04-28)
This work was motivated by the need of stimuli responsive drug carriers, which can be activated by low cost non-invasive stimuli such as external magnetic field (EMF). Thus, novel antimicrobial materials based on xanthan gum (XG), magnetic nanoparticles (MNP), bovine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service