Skip to Content
Merck
All Photos(1)

Key Documents

348279

Sigma-Aldrich

Diallyldimethylammonium chloride solution

65 wt. % in H2O

Synonym(s):

Dimethyldiallylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N+(CH2CH=CH2)2]Cl-
CAS Number:
Molecular Weight:
161.67
Beilstein:
5830817
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

concentration

65 wt. % in H2O

SMILES string

[Cl-].C[N+](C)(CC=C)CC=C

InChI

1S/C8H16N.ClH/c1-5-7-9(3,4)8-6-2;/h5-6H,1-2,7-8H2,3-4H3;1H/q+1;/p-1

InChI key

GQOKIYDTHHZSCJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Diallyldimethylammonium chloride solution (DADMAC) is a hydrophilic quaternary ammonium compound that can be dissolved in an aqueous solution as a positively charged colloid.

Application

DADMAC is used as a cationic monomer solution for the fabrication of ion-selective polyelectrolytic anodized aluminium oxide (AAO) membranes which can be used for the development of electrical power generation systems. It may be grafted on carboxymethyl cellulose (CMC) for use as an absorbent for cationic dyes.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiwall carbon nanotubes-poly (diallyldimethylammonium chloride)-graphene hybrid composite film for simultaneous determination of catechol and hydroquinone.
Song D, et al.
Sensors and Actuators B, Chemical, 206(1), 111-118 (2015)
The mining chemical Polydadmac is cytotoxic but does not interfere with Cu-induced toxicity in Atlantic salmon hepatocytes.
Olsvik P, et al.
Toxicology in vitro, 30(1), 492-505 (2015)
Adsorption properties of crosslinking carboxymethyl cellulose grafting dimethyldiallylammonium chloride for cationic and anionic dyes.
Lin Q, et al.
Carbohydrate Polymers, 151(1), 283-294 (2016)
Densely charged polyelectrolyte-stuffed nanochannel arrays for power generation from salinity gradient.
Kwak H, et al.
Scientific Reports, 6(1), 26416-26416 (2016)
Andre Jung et al.
Soft matter, 14(40), 8105-8111 (2018-08-02)
Highly alkaline hydrogels that allow the restoration of alkaline buffer in cementitious materials can be obtained from diallyldimethylammonium hydroxide. The latter must be prepared in dilute solutions and polymerised at ambient temperatures in order to avoid decomposition. Using methacrylamide as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service