Skip to Content
Merck
All Photos(2)

Key Documents

C109800

Sigma-Aldrich

Cyclooctanone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H14(=O)
CAS Number:
Molecular Weight:
126.20
Beilstein:
1280738
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

bp

195-197 °C (lit.)

mp

32-41 °C (lit.)

density

0.958 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCCCC1

InChI

1S/C8H14O/c9-8-6-4-2-1-3-5-7-8/h1-7H2

InChI key

IIRFCWANHMSDCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

165.2 °F

Flash Point(C)

74 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vishwakarma Singh et al.
The Journal of organic chemistry, 70(3), 973-981 (2005-01-29)
A new and efficient synthesis of a variety of highly embellished bicyclooctenones having an endo-vinyl moiety and their sigmatropic shifts in ground and excited states leading to a stereoselective route to substituted cis-decalins and diquinane frameworks have been described. Functionalized
K Yamada et al.
Chemical & pharmaceutical bulletin, 45(12), 1898-1905 (1998-01-20)
Construction of the AB-ring system of the taxane framework via an A-ring annulation strategy was demonstrated by base-mediated intramolecular aldol reaction of (Z)-2,2-dimethyl-3-(1-methyl-2-oxopropylidene)cyclooctanone, affording the title compound, 1-hydroxy-8,11,11-trimethylbicyclo[5.3.1]undec-7-en-9-one. A cyclization precursor, the tetra-substituted (Z)-alkene, was prepared from the corresponding cyclooctanone
F E Harvey et al.
Brain research bulletin, 13(4), 541-547 (1984-10-01)
Female mice were reared in observation incubators from day 1 of life for three weeks. During that time they were continuously exposed to the odors of either cyclooctanone, adult male mouse urine or distilled water. The growth rate was temporarily
M E Krafft et al.
The Journal of organic chemistry, 66(22), 7443-7448 (2001-10-30)
The total synthesis of asteriscanolide (1) has been achieved by taking advantage on an intermolecular Pauson-Khand cycloaddition and a ring-closing metathesis as key bond-forming transformations. The approach incorporates the cyclooctane stereogenic center prior to ring formation. Interestingly, the ring-closing metathesis
Korany A Ali et al.
Archiv der Pharmazie, 345(3), 231-239 (2011-11-03)
The versatile synthon (E)-2-((dimethyl amino)methylene)cyclooctanone (2) was used as a key intermediate for the synthesis of cyclooctanones and cyclooctane-based heterocycles with pyrazole, isoxazole, pyrimidine, pyrazolopyrimidine, triazolopyrimidine and imidazopyrimidine derivatives via its reactions with several nitrogen nucleophiles. The newly synthesized compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service