Skip to Content
Merck
All Photos(3)

Key Documents

234192

Sigma-Aldrich

1-Hexanethiol

95%

Synonym(s):

1-Mercaptohexane, Hexyl mercaptan, MCH, Mercaptan C6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5SH
CAS Number:
Molecular Weight:
118.24
Beilstein:
1731295
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

95%

form

liquid

refractive index

n20/D 1.450

bp

150-154 °C (lit.)

mp

−81-−80 °C (lit.)

density

0.832 g/mL at 25 °C

SMILES string

CCCCCCS

InChI

1S/C6H14S/c1-2-3-4-5-6-7/h7H,2-6H2,1H3

InChI key

PMBXCGGQNSVESQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hexanethiol is an organothiol that forms a self-assembled monolayer (SAM) on a variety of nanoparticles. It consists of S head group, CH2 alkane chain, and CH3 terminal group.[1][2]

Application

1-Hexanethiol can be mixed with 4-mercaptophenol to functionalize gold nanoparticles (AuNPs) for use as chemi-resistive sensors.[3] It can also form a protective coating against oxidative reactions and contaminating layers on copper surfaces.[2] It can also be used as a thiol based ligand which finds potential application as a semiconducting material in thin film solar cells.[4]
It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.[5] It is also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.[6]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noble metal surface degradation induced by organothiols
de Poel W, et al.
Surface Science, 662, 59-66 (2017)
T Hashimoto et al.
The British journal of dermatology, 134(2), 336-339 (1996-02-01)
In this study we present a patient with the sublamina densa type of linear IgA bullous dermatosis (LABD), with IgA autoantibodies reactive with the 290-kDa type VII collagen (the epidermolysis bullosa acquisita (EBA) antigen) and with immunoblotting of normal human
Tan, C. S.; Lim, D. F.; Singh, S. G.; Goulet, S. K.; Bergkvist, M.
Applied Physics Letters, 91, 192108-192108 (2007)
Different ligand exchange solvents effect on the densification of CuIn0. 7Ga0. 3Se2 prepared using the heating-up method
Yang C and Hsiang H
Applied Surface Science, 426(2), 1148-1157 (2017)
N Yasaka et al.
Journal of dermatological science, 7(2), 136-141 (1994-04-01)
The cutaneous basement membrane zone (BMZ) is composed of a large number of molecular components. We have recently reported that one of the CD1b monoclonal antibodies (MoAb), NU-T2, reacts to the BMZ of the normal human skin, esophagus and stomach.

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service