Skip to Content
Merck
All Photos(3)

Documents

C102504

Sigma-Aldrich

Cyclohexene oxide

98%

Synonym(s):

1,2-Epoxycyclohexane, 7-Oxabicyclo[4.1.0]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O
CAS Number:
Molecular Weight:
98.14
Beilstein:
383568
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid

autoignition temp.

703 °F

expl. lim.

12.36 %

refractive index

n20/D 1.452 (lit.)

bp

129-130 °C (lit.)

density

0.97 g/mL at 25 °C (lit.)

SMILES string

C1CCC2OC2C1

InChI

1S/C6H10O/c1-2-4-6-5(3-1)7-6/h5-6H,1-4H2

InChI key

ZWAJLVLEBYIOTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Polymeric carbon nitride with internal np homojunctions for efficient photocatalytic CO2 reduction coupled with cyclohexene oxidation: This study focuses on the use of cyclohexene oxide in the context of photocatalytic CO2 reduction, highlighting the application of polymeric carbon nitride as a catalyst. The process shows how cyclohexene oxide can be efficiently converted in a coupled reaction that also addresses environmental concerns through CO2 reduction (W Zhen et al., 2021).

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathila S Rajapaksa et al.
Toxicological sciences : an official journal of the Society of Toxicology, 96(2), 327-334 (2007-01-06)
Ovarian follicle disruption in mice caused by 7,12-dimethylbenz[a]anthracene (DMBA) is attributed to its bioactivation by CYP1B1 to a 3,4-epoxide which is then hydrolyzed to form a 3,4-diol by microsomal epoxide hydrolase (mEH). Further epoxidation by CYP1A1 or 1B1 forms the
Alternating copolymerization of epoxides and cyclic anhydrides: an improved route to aliphatic polyesters.
Ryan C Jeske et al.
Journal of the American Chemical Society, 129(37), 11330-11331 (2007-08-29)
Youli Xiao et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 11(12), 3668-3678 (2005-04-14)
The intramolecular dinuclear zinc complexes generated in situ from the reaction of multidentate semi-azacrown ether ligands with Et(2)Zn, followed by treatment with an alcohol additive, were found to promote the copolymerization of CO(2) and cyclohexene oxide (CHO) with completely alternating
Donald J Darensbourg et al.
Journal of the American Chemical Society, 127(40), 14026-14038 (2005-10-06)
The mechanism of the copolymerization of cyclohexene oxide and carbon dioxide to afford poly(cyclohexylene)carbonate catalyzed by (salen)CrN3 (H2salen = N,N,'-bis(3,5-di-tert-butylsalicylidene)-1,2-ethylene-diimine) in the presence of a broad range of cocatalysts has been studied. We have previously established the rate of copolymer
Highly active dizinc catalyst for the copolymerization of carbon dioxide and cyclohexene oxide at one atmosphere pressure.
Michael R Kember et al.
Angewandte Chemie (International ed. in English), 48(5), 931-933 (2008-12-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service