BR128954
BRAND® narrow-mouth bottles, PFA
capacity 500 mL
Synonym(s):
Bottles, PFA, Plastics
About This Item
Recommended Products
material
PFA bottle
PFA cap
clear bottle
narrow-mouth bottle
round bottle
Joint
Joints threaded neck (S28)
feature
autoclavable
with cap
packaging
pack of 1 ea
manufacturer/tradename
BRAND 128954
capacity
500 mL
diam. × H
76 mm × 190 mm
General description
Bottles are designed for long-term storage of solvents and standards (trace analytics). PFA screw cap with a molded sealing ring for a positive seal. Autoclavable, easy to clean, withstands temperatures from -200 °C to 260 °C.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)