Skip to Content
Merck
All Photos(2)

Documents

B7149

Sigma-Aldrich

Benzthiazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14ClN3O4S3
CAS Number:
Molecular Weight:
431.94
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

SMILES string

NS(=O)(=O)c1cc2c(NC(CSCc3ccccc3)=NS2(=O)=O)cc1Cl

InChI

1S/C15H14ClN3O4S3/c16-11-6-12-14(7-13(11)25(17,20)21)26(22,23)19-15(18-12)9-24-8-10-4-2-1-3-5-10/h1-7H,8-9H2,(H,18,19)(H2,17,20,21)

InChI key

NDTSRXAMMQDVSW-UHFFFAOYSA-N

Gene Information

human ... SLC12A3(6559)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a productā€™s label following the words ā€˜Lotā€™ or ā€˜Batchā€™.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C Meyer et al.
Biopharmaceutics & drug disposition, 3(1), 1-9 (1982-01-01)
An assay was developed for benzthiazide in plasma, urine and feces, using high performance liquid chromatography (HPLC). A reverse-phase column was employed, with quantitation af 280 nm, using polythiazide as an internal standard. In three of four human subjects who
Thiazides XIII: Dissolution testing: a bioavailability predictor for benzthiazide tablets.
V P Shah et al.
Biopharmaceutics & drug disposition, 3(3), 283-285 (1982-07-01)
J P Huang et al.
Journal of analytical toxicology, 23(5), 337-342 (1999-09-17)
This study describes a matrix-assisted laser-desorption-ionization (MALDI) mass spectrometry for rapid screening of 12 diuretics in spiked urine. C60 is used as the matrix for MALDI. Diuretics are directly analyzed by C60-MALDI without previous derivatization. The fact that most diuretic
Laurie De Wilde et al.
Journal of chromatography. A, 1579, 31-40 (2018-11-16)
Diuretics can be misused to force diuresis to achieve weight loss or to mask the intake of a prohibited substance and are therefore prohibited by the World Anti-Doping Agency (WADA). For similar reasons other masking agents (vaptans, probenecid, etc.) are
L R Berkowitz
The American journal of physiology, 258(4 Pt 1), C622-C629 (1990-04-01)
The thioalkylating agent N-ethylmaleimide (NEM) causes ouabain-insensitive K loss from human red blood cells. This K loss is inhibited when intracellular Cl is replaced by another permeant anion or when loop diuretics are placed in the incubation medium after NEM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service