Skip to Content
Merck

A4438

p-Acetamidophenyl β-D-glucuronide sodium salt

>98% (TLC)

Synonym(s):

Acetaminophen glucuronide, Paracetamol glucuronide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C14H16NO8Na
CAS Number:
Molecular Weight:
349.27
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

>98% (TLC)

form

powder

impurities

<6% water (Karl Fischer)

color

white to off-white

solubility

water: 25 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

[Na+].CC(=O)Nc1ccc(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C([O-])=O)cc1

InChI

1S/C14H17NO8.Na/c1-6(16)15-7-2-4-8(5-3-7)22-14-11(19)9(17)10(18)12(23-14)13(20)21;/h2-5,9-12,14,17-19H,1H3,(H,15,16)(H,20,21);/q;+1/p-1/t9-,10-,11+,12-,14+;/m0./s1

InChI key

OINXIJJEOMGKPB-CYRSAHDMSA-M

Application

Acetaminophen glucuronide (AA-Gluc) is used as a reference material in procedures that analyse urine and plasma for acetaminophen metabolites to indicate hepatotoxicity.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



T Andrew Clayton et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(34), 14728-14733 (2009-08-12)
We provide a demonstration in humans of the principle of pharmacometabonomics by showing a clear connection between an individual's metabolic phenotype, in the form of a predose urinary metabolite profile, and the metabolic fate of a standard dose of the
Scott A Reisman et al.
Toxicological sciences : an official journal of the Society of Toxicology, 109(1), 31-40 (2009-02-28)
Acetaminophen (AA) is a widely used antipyretic drug that causes hepatotoxicity at high doses. Nuclear factor erythroid 2-related factor 2 (Nrf2) is a transcription factor that mitigates electrophilic stress from AA by inducing genes, such as NAD(P)H:quinone oxidoreductase 1 (Nqo1)
Carol R Gardner et al.
Toxicological sciences : an official journal of the Society of Toxicology, 125(2), 607-612 (2011-11-04)
Fenbendazole is a broad-spectrum anthelmintic drug widely used to prevent or treat nematode infections in laboratory rodent colonies. Potential interactions between fenbendazole and hepatotoxicants such as acetaminophen are unknown, and this was investigated in this study. Mice were fed a



Global Trade Item Number

SKUGTIN
A4438-10MG04061832386942
A4438-50MG04061832386959