Skip to Content
Merck
All Photos(2)

Key Documents

ALD00006

Sigma-Aldrich

Sodium bis(2-hydroxy-2-methylbutyrato)oxochromate(V)

Synonym(s):

2-Hydroxy-2-methyl-butanoic acid chromium complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16CrNaO7
CAS Number:
Molecular Weight:
323.22
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

340-350 °C

SMILES string

[O-][Cr]1(O2)(OC(C(C)(CC)O1)=O)OC(C)(CC)C2=O.[Na+]

InChI

1S/2C5H9O3.Cr.Na.O/c2*1-3-5(2,8)4(6)7;;;/h2*3H2,1-2H3,(H,6,7);;;/q2*-1;+4;+1;-1/p-2

InChI key

NIZKSTMLMALTGU-UHFFFAOYSA-L

Application

Oxidant was reported by the Baran group to promote selective allylic oxidation of a synthetic intermediate derived from (+)-taxadiene en route to the synthesis of (−)-taxyunnanine D.

Two-Phase Synthesis of (−)-Taxuyunnanine D

Pictograms

Flame over circle

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service