Skip to Content
Merck
All Photos(1)

Key Documents

8.12382

Sigma-Aldrich

Titanium(IV) chloride

for synthesis

Synonym(s):

Titanium(IV) chloride, Titanium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl4Ti
CAS Number:
Molecular Weight:
189.68
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-441-9
NACRES:
NA.22
form:
liquid

vapor pressure

12 hPa ( 20 °C)

Quality Level

form

liquid

potency

3160 mg/kg LD50, skin (Rabbit)

bp

136 °C/1013 hPa

mp

-24 °C

density

1.728 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Ti+4].[Cl-].[Cl-].[Cl-].[Cl-]

InChI

1S/4ClH.Ti/h4*1H;/q;;;;+4/p-4

InChI key

XJDNKRIXUMDJCW-UHFFFAOYSA-J

Application

Titanium(IV) chloride can be used as a Lewis acid catalyst:
  • In the ortho acylation of aryl alcohols to synthesize corresponding ortho C-acylated products.
  • To synthesize various chlorinated compounds via Baylis−Hillman reaction reactions of aryl aldehydes with methyl vinyl ketone in the presence of Lewis base.
  • To prepare 3,4-dihydropyrimidin-2(1H)-ones and thiones by Biginelli condensation reaction of aldehyde, 1,3-dicarbonyl compound, and urea or thiourea.
  • In the hydrodefluorination reaction in the presence of triethylsilane as the reducing agent.

Analysis Note

Assay (argentometric): ≥ 97.0 %
Identity (ICP-OES): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transformation of Trifluorotoluenes Triggered by Titanium (IV) Chloride-Catalyzed Hydrodefluorination using Hydrosilanes
Yamada T, et al.
Advanced Synthesis & Catalysis, 358(1), 62-66 (2016)
Transformation of Trifluorotoluenes Triggered by Titanium (IV) Chloride-Catalyzed Hydrodefluorination using Hydrosilanes
Yamada T, et al.
advanced synthesis and catalysis, 358(1), 62-66 (2016)
Titanium (IV) chloride-mediated ortho-acylation of phenols and naphthols
Bensari A and Zaveri NT
Synthesis, 2003(02), 0267-0271 (2003)
Titanium (IV) chloride catalyzed one-pot synthesis of 3, 4-dihydropyrimidin-2 (1H)-ones and thiones
Nagawade RR, et al.
Mendeleev Communications, 15(4), 150-151 (2005)
Titanium(IV) chloride and the amine-promoted baylis-hillman reaction
M Shi, et al.
Organic Letters, 2(16), 2397-2400 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service