Skip to Content
Merck
All Photos(2)

Key Documents

A76109

Sigma-Aldrich

3-Amino-1-propanesulfonic acid

97%

Synonym(s):

3APS, Homotaurine, Tramiprosate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)3SO3H
CAS Number:
Molecular Weight:
139.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

293 °C (dec.) (lit.)

SMILES string

NCCCS(O)(=O)=O

InChI

1S/C3H9NO3S/c4-2-1-3-8(5,6)7/h1-4H2,(H,5,6,7)

InChI key

SNKZJIOFVMKAOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Important gabamimetic agent-evokes neurological responses similar to those elicited by γ-aminobutyric acid (GABA).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francine Gervais et al.
Neurobiology of aging, 28(4), 537-547 (2006-05-06)
Amyloid beta-peptide (Abeta) is a major constituent of senile plaques in Alzheimer's disease (AD). Neurotoxicity results from the conformational transition of Abeta from random-coil to beta-sheet and its oligomerization. Among a series of ionic compounds able to interact with soluble
Eric Martineau et al.
Journal of the American Society for Mass Spectrometry, 21(9), 1506-1514 (2010-06-29)
This paper describes an efficient and reproducible screening method for identifying low molecular weight compounds that bind to amyloid beta peptides (Abeta) peptides using electrospray ionization mass spectrometry (ESI-MS). Low molecular weight compounds capable of interacting with soluble Abeta may
Steven M Greenberg et al.
Alzheimer disease and associated disorders, 20(4), 269-274 (2006-11-30)
No treatments have been identified to lower the risk of intracerebral hemorrhage due to cerebral amyloid angiopathy (CAA). A potential approach to prevention is the use of agents that interfere with the pathogenic cascade initiated by the beta-amyloid peptide (Abeta).
Xiao-Dong Hu et al.
Neuropharmacology, 50(2), 238-244 (2005-12-06)
The benzodiazepine diazepam impairs memory and long-term potentiation (LTP) in the hippocampus. Here, we investigate the effect of diazepam on LTP of C-fiber evoked field potentials in spinal dorsal horn, which is relevant to pathological pain. LTP of C-fiber evoked
M Foster Olive et al.
European journal of pharmacology, 437(1-2), 55-61 (2002-02-28)
The purpose of the present study was to determine the acute effects of the anticraving compound acamprosate (calcium acetylhomotaurinate) and the closely related compound homotaurine on ethanol intake and ethanol-stimulated dopamine release in the nucleus accumbens. Male rats were treated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service