389803
3-Hydroxy-1-propanesulfonic acid sodium salt
technical grade, 80%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HO(CH2)3SO3Na
CAS Number:
Molecular Weight:
162.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
grade
technical grade
Quality Level
Assay
80%
mp
260 °C (dec.) (lit.)
functional group
hydroxyl
sulfonic acid
SMILES string
[Na+].OCCCS([O-])(=O)=O
InChI
1S/C3H8O4S.Na/c4-2-1-3-8(5,6)7;/h4H,1-3H2,(H,5,6,7);/q;+1/p-1
InChI key
CSKVLUWCGPWCQR-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service