Skip to Content
Merck
All Photos(1)

Key Documents

A15401

Sigma-Aldrich

Acetylenedicarboxylic acid monopotassium salt

≥98%

Synonym(s):

mono-Potassium 2-butynedioate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCC≡CCOOK
CAS Number:
Molecular Weight:
152.15
Beilstein:
3571639
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

powder

SMILES string

[K+].OC(=O)C#CC([O-])=O

InChI

1S/C4H2O4.K/c5-3(6)1-2-4(7)8;/h(H,5,6)(H,7,8);/q;+1/p-1

InChI key

KLLYWRUTRAFSJT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of an intermediate in the de novo formation of nicotinamide adenine dinucleotide in Escherichia coli.
J T Heard et al.
Biochemical and biophysical research communications, 95(4), 1517-1521 (1980-08-29)
Lothar Brecker et al.
European journal of biochemistry, 270(7), 1393-1398 (2003-03-26)
Only 2% of the known natural products with acetylenic bonds are alpha-alkynoates. Their polarized, conjugated triple bond is an optimal target for an enzymic hydration. Therefore they are good substrates for the enzymes involved in metabolism of acetylenic compounds, resulting
A S Subbaraman et al.
Origins of life, 10(4), 343-347 (1980-12-01)
A number of routes have been suggested for the prebiotic synthesis of uracil involving the reaction of urea with malic acid, propiolic acid, cyanoacetylene and others. Cyanoacetylene has been detected in the interstellar medium as well as simulated prebiotic experiments.
Issa Yavari et al.
Molecular diversity, 10(2), 247-250 (2006-06-14)
The adduct produced in the reaction between alkyl isocyanides and dialkyl acetylenedicarboxylates was trapped by 2,2,5-trimethyl-1,3-dioxane-4,6-dione (methyl Meldrum's acid), to afford highly functionalized ketenimines in good yields.
Kyungho Park et al.
The Journal of organic chemistry, 75(18), 6244-6251 (2010-08-28)
Symmetrical diarylalkynes were obtained from propiolic acid (or 2-butynedioic acid) and aryl halides in good yields. The optimized reaction conditions were 2.0 equiv of aryl halide, 1.0 equiv of propiolic acid, 5.0 mol % Pd(PPh(3))(2)Cl(2), 10.0 mol % 1,4-bis(diphenylphosphino)butane (dppb)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service