All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H3NO2
CAS Number:
Molecular Weight:
97.07
Beilstein:
106910
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
powder
mp
91-93 °C (lit.)
functional group
maleimide
SMILES string
O=C1NC(=O)C=C1
InChI
1S/C4H3NO2/c6-3-1-2-4(7)5-3/h1-2H,(H,5,6,7)
InChI key
PEEHTFAAVSWFBL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Maleimide (2,5-Pyrroledione) is a new nanoparticle surface functional group which favors easy conjugation with cell penetration peptides. The conjugation is enabled via click chemistry to preserve its biofunctions.
Maleimide is used as a dienophile in Diels Alder reaction for the synthesis of macromolecules.
Maleimide is used as a dienophile in Diels Alder reaction for the synthesis of macromolecules.
Application
Rhodium-catalyzed conjugate arylation with arylboronic acids.
The maleimide-functionalized silica beads were used to immobilize the bovine serum albumin (BSA)-boronic acid (BA) conjugates.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Guangxue Feng et al.
Small (Weinheim an der Bergstrasse, Germany), 10(6), 1212-1219 (2013-12-18)
Single-chain conjugated polymer (CP) dots embedded nanoparticles (NPs) bearing cell penetration peptide (TAT) as surface ligands are synthesized for long term cancer cell tracing applications. The CPNPs are fabricated by matrix-encapsulation method and the embedded CPs can be modulated into
Paulo L Onuchic et al.
Scientific reports, 9(1), 12161-12161 (2019-08-23)
Liquid-liquid phase separation (LLPS) of RNA-protein complexes plays a major role in the cellular function of membraneless organelles (MLOs). MLOs are sensitive to changes in cellular conditions, such as fluctuations in cytoplasmic ion concentrations. To investigate the effect of these
Satya Nandana Narla et al.
Biochemical and biophysical research communications, 443(2), 562-567 (2013-12-12)
We report bovine serum albumin (BSA)-boronic acid (BA) conjugates as lectin mimetics and their glyco-capturing capacity. The BSA-BA conjugates were synthesized by amidation of carboxylic acid groups in BSA with aminophenyl boronic acid in the presence of EDC, and were
Meng Ke et al.
PLoS computational biology, 13(6), e1005603-e1005603 (2017-06-16)
GLUT1 facilitates the down-gradient translocation of D-glucose across cell membrane in mammals. XylE, an Escherichia coli homolog of GLUT1, utilizes proton gradient as an energy source to drive uphill D-xylose transport. Previous studies of XylE and GLUT1 suggest that the
The furan/maleimide Diels--Alder reaction: A versatile click--unclick tool in macromolecular synthesis
Gandini A.
Progress in Polymer Science, 38, 1-29 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service