Skip to Content
Merck
All Photos(1)

Documents

96410

Sigma-Aldrich

Methyl cinnamate

≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOOCH3
CAS Number:
Molecular Weight:
162.19
Beilstein:
386468
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (GC)

form

crystals

bp

260-262 °C (lit.)

mp

33-38 °C (lit.)

SMILES string

COC(=O)\C=C\c1ccccc1

InChI

1S/C10H10O2/c1-12-10(11)8-7-9-5-3-2-4-6-9/h2-8H,1H3/b8-7+

InChI key

CCRCUPLGCSFEDV-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathrin Fink et al.
Journal of agricultural and food chemistry, 52(10), 3065-3068 (2004-05-13)
For the authenticity assessment of (E)-methyl cinnamate from different origins, combustion/pyrolysis-isotope ratio mass spectrometry (C/P-IRMS) was used by an elemental analyzer (EA) and on-line capillary gas chromatography coupling (HRGC-C/P-IRMS). For that reason, (E)-methyl cinnamate self-prepared from synthetic, natural, and semisynthetic
S P Bhatia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45 Suppl 1, S113-S119 (2007-11-27)
A toxicologic and dermatologic review of methyl cinnamate when used as a fragrance ingredient is presented.
Bhanu Prakash et al.
International journal of food microbiology, 153(1-2), 183-191 (2011-12-06)
The investigation deals with antifungal, antiaflatoxin and antioxidant efficacy of Zanthoxylum alatum Roxb. essential oil (EO), its two major constituents and their comparison with five commonly used organic acid preservatives. The chemical profile of EO, characterized through GC and GC-MS
Yun-Yu Chen et al.
Journal of agricultural and food chemistry, 60(4), 955-963 (2012-01-26)
Methyl cinnamate, an active component of Zanthoxylum armatum , is a widely used natural flavor compound with antimicrobial and tyrosinase inhibitor activities. However, the underlying bioactivity and molecular mechanisms of methyl cinnamate on adipocyte function and metabolism remain unclear. The
L P Delbressin et al.
Archives of toxicology, 49(3-4), 321-330 (1982-03-01)
The urinary mercapturic acid excretion by female rats of methyl atropate (alpha-phenyl methyl acrylate) and methyl cinnamate (beta-phenyl methyl acrylate) has been studied. On the basis of the structures of these mercapturic acids the conclusion can be drawn that these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service