Skip to Content
Merck
All Photos(3)

Documents

S4972

Sigma-Aldrich

Styrene

ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%

Synonym(s):

Phenylethylene, Vinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CH2
CAS Number:
Molecular Weight:
104.15
Beilstein:
1071236
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.6 (vs air)

Quality Level

vapor pressure

12.4 mmHg ( 37.7 °C)
4.3 mmHg ( 15 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

914 °F

contains

4-tert-butylcatechol as stabilizer

expl. lim.

6.1 %

refractive index

n20/D 1.546 (lit.)

bp

145-146 °C (lit.)

mp

−31 °C (lit.)

density

0.906 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C=Cc1ccccc1

InChI

1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2

InChI key

PPBRXRYQALVLMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Removal of styrene from textile dyeing wastewater treatment plants (TDWTPs) has been investigated. Catalytic potential of the Co-SBA-15 for the oxidation of styrene with hydrogen peroxide has been investigated. Screening of asymmetric epoxidation of styrene by gas chromatography-mass spectrometry (GC-MS) with selected ion monitoring (SIM) has been reported. The homogeneous atom transfer radical polymerization (ATRP) of styrene by employing solubilizing 4,4′-dialkyl substituted 2,2′-bipyridines affords well-defined polymers. The Heck reaction of chlorobenzene and substituted chlorobenzenes with styrene in water using catalytic palladium on carbon affords 1,2-diphenylethane.

Application

Styrene has been used as a test styrene sample in a study involving the effect of reversible addition-fragmentation chain transfer (RAFT) controllers on its RAFT polymerization in supercritical carbon dioxide in the presence of poly(styrene-block-dimethylsiloxane) (PSDMS) as a stabilizer.
Styrene was used in the preparation of well-defined homopolymers, diblock copolymers and triblock copolymers of tert-butyl acrylate, methyl acrylate and styrene.

Other Notes

Inhibitor depletes over time especially at higher temperatures

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

hearing organs, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heterogeneous palladium-catalysed heck reaction of aryl chlorides and styrene in water under mild conditions.
Mukhopadhyay S, et al.
Advanced Synthesis & Catalysis, 344(3-4), 348-354 (2002)
Xun-An Ning et al.
Chemosphere, 136, 50-55 (2015-05-02)
The occurrence and removal of benzene, toluene, ethylbenzene, xylenes, styrene and isopropylbenzene (BTEXSI) from 6 textile dyeing wastewater treatment plants (TDWTPs) were investigated in this study. The practical capacities of the 6 representative plants, which used the activated sludge process
Effect of stabilizer concentration and controller structure and composition on polymerization rate and molecular weight development in RAFT polymerization of styrene in supercritical carbon dioxide.
Jaramillo-Soto G, et al.
Polymer, 50(21), 5024-5030 (2009)
The preparation of t-butyl acrylate, methyl acrylate, and styrene block copolymers by atom transfer radical polymerization: Precursors to amphiphilic and hydrophilic block copolymers and conversion to complex nanostructured materials.
Ma Q and Wooley KL.
Journal of Polymer Science Part A: Polymer Chemistry, 38(S1), 4805-4820 (2000)
Xiaojing Jin et al.
Journal of nanoscience and nanotechnology, 14(9), 7234-7241 (2015-05-01)
The rapid preparation of SBA-15 with cobalt (Co) introduction was performed via the ultrasonic irradiation in combination with "pH-adjusting" method. The catalytic properties of the synthesized Co-SBA-15 were investigated by examining the oxidation of styrene with hydrogen peroxide. The effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service