Skip to Content
Merck
All Photos(2)

Key Documents

934003

Sigma-Aldrich

Methyl (2,2,2-trifluoroethyl) carbonate

greener alternative

≥98%, anhydrous, battery grade

Synonym(s):

Carbonic Acid Methyl 2,2,2-Trifluoroethyl Ester, FEMC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5F3O3
CAS Number:
Molecular Weight:
158.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

battery grade

Quality Level

Assay

≥98%

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤250 ppm HF
≤300 ppm H2O

bp

57-59 °C/120 mmHg
90 °C (lit.)

density

1.34 g/mL (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

FC(F)(F)COC(=O)OC

InChI key

GBPVMEKUJUKTBA-UHFFFAOYSA-N

General description

Methyl (2,2,2-trifluoroethyl) carbonate (FEMC), ≥99%, battery grade is one kind of fluorinated linear carbonate. It comes with very low water content (≤ 300.0 ppm) and low impurity of HF (≤ 250.0 ppm) which is ideal for battery research
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Battery-grade methyl (2,2,2-trifluoroethyl) carbonate (FEMC) is an effective additive for improving the performance and stability of lithium-ion batteries. Its outstanding thermal and electrochemical stability makes it a reliable option for high-temperature and high-voltage battery applications. FEMC has been shown to improve battery life, enhance energy density, and increase cycling stability. As an electrolyte additive in lithium-ion batteries (LIBs), FEMC plays an important role in forming a stable solid electrolyte interphase (SEI) and effective passivation of the cathode surfaces, resulting in enhanced discharge capacity and cyclability. Even the addition of 5 wt % of FEMC was found to be very effective in forming a stable SEI layer and protecting the surface of the cathode. FEMC′s high performance makes it an attractive choice for battery manufacturers looking to create safe and efficient battery products.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F

Flash Point(C)

38 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Building Safe Lithium-Ion Batteries for Electric Vehicles: A Review
Duan J, et al.
Electrochemical Energy Reviews, 3, 1-42 (2020)
Interfacial Origin of Performance Improvement and Fade for 4.6 V. LiNi0.5Co0.2Mn0.3O2 Battery Cathodes
Lee Yu-Mi, et al.
The Journal of Physical Chemistry C, 118, 10631?10639-10631?10639 (2014)
Fluorinated Carbonate-Based Electrolyte for High-Voltage Li(Ni0.5Mn0.3Co0.2)O2/Graphite Lithium-Ion Battery
Jinsol Im, et al.
Journal of the Electrochemical Society, 164 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service