Skip to Content
Merck
All Photos(1)

Key Documents

931861

Sigma-Aldrich

(3aR,3′aR,8aS,8′a′S)-2,2′-Cyclopropylidenebis[3a,8a-dihydro-8H-Indeno[1,2-d]oxazole]

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20N2O2
CAS Number:
Molecular Weight:
356.42
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

SMILES string

N1=C(O[C@@H]6[C@H]1c7c(cccc7)C6)C5(CC5)C2=N[C@H]3[C@@H](O2)Cc4c3cccc4

InChI

1S/C23H20N2O2/c1-3-7-15-13(5-1)11-17-19(15)24-21(26-17)23(9-10-23)22-25-20-16-8-4-2-6-14(16)12-18(20)27-22/h1-8,17-20H,9-12H2/t17-,18-,19+,20+/m0/s1

InChI key

QFXUVUBVPPOPQZ-VNTMZGSJSA-N

Application

(3aR,3′aR,8aS,8′a′S)-2,2′-Cyclopropylidenebis[3a,8a-dihydro-8H-Indeno[1,2-d]oxazole] is a bisoxazoline (BOX) ligand for asymmetric catalysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper(I)-catalyzed highly enantioselective [3+3]-cycloaddition of ?-alkyl enoldiazoacetates with nitrones
Dong K, et al.
Organic Chemistry Frontiers : An International Journal of Organic Chemistry / Royal Society of Chemistry, 7, 1653-1657 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service