Skip to Content
Merck
All Photos(1)

Key Documents

92748

Sigma-Aldrich

4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate

≥97.0%, for peptide synthesis

Synonym(s):

2-(Trimethylsilyl)ethyl 4-nitrophenyl carbonate, TEOC-ONp

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO5Si
CAS Number:
Molecular Weight:
283.35
Beilstein:
4320585
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Product Name

4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate, ≥97.0%

Quality Level

Assay

≥97.0%

form

crystals

mp

35-40 °C

application(s)

peptide synthesis

functional group

carbonate
nitro

storage temp.

2-8°C

SMILES string

C[Si](C)(C)CCOC(=O)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C12H17NO5Si/c1-19(2,3)9-8-17-12(14)18-11-6-4-10(5-7-11)13(15)16/h4-7H,8-9H2,1-3H3

InChI key

ZAQWGGKIMQIVGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate can be used as:
  • A protecting group for an amine in one of the key synthetic steps of kottamide E total synthesis.
  • A starting material to prepare a cis-butene derivative, which is used as a polymer terminating agent in the synthesis of monotelechelic glycopolymers.

Other Notes

Reagent for the introduction of the TEOC-amino protecting group which can be cleaved by F-

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A. Rosowsky et al.
The Journal of Organic Chemistry, 54, 5551-5551 (1989)
End-labeled amino terminated monotelechelic glycopolymers generated by ROMP and Cu (I)-catalyzed azide--alkyne cycloaddition
Okoth R and Basu A
Beilstein Journal of Organic Chemistry, 9(1), 608-612 (2013)
B Wünsch et al.
Hoppe-Seyler's Zeitschrift fur physiologische Chemie, 362(9), 1289-1292 (1981-09-01)
2-(Trimethylsilyl)ethyl-4-nitrophenyl carbonate has been prepared as a new reagent for the introduction of the 2-(trimethylsilyl)ethyloxycarbonyl group into amino acids or amino acid derivatives. The resulting N alpha-protected amino acids were found to represent suitable intermediates for the synthesis of peptides.
L.A. Carpino et al.
Journal of the Chemical Society. Chemical Communications, 358-358 (1978)
Thomas B Parsons et al.
Chemical communications (Cambridge, England), 49(23), 2296-2298 (2013-02-12)
The first synthesis of kottamide E, a marine natural product containing a 5,6-dibromoindole linked via a (Z)-enamide to an unusual 1,2-dithiolane-containing amino acid, is reported.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service