Skip to Content
Merck
All Photos(4)

Documents

160210

Sigma-Aldrich

4-Nitrophenyl chloroformate

96%, for peptide synthesis

Synonym(s):

Chloroformic acid 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO2C6H4NO2
CAS Number:
Molecular Weight:
201.56
Beilstein:
518127
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

product name

4-Nitrophenyl chloroformate, 96%

Assay

96%

form

powder

reaction suitability

reaction type: Carbonylations

bp

159-162 °C/19 mmHg (lit.)

mp

77-79 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(OC(Cl)=O)cc1

InChI

1S/C7H4ClNO4/c8-7(10)13-6-3-1-5(2-4-6)9(11)12/h1-4H

InChI key

NXLNNXIXOYSCMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Nitrophenyl chloroformate, also known as p-nitrophenyl chloroformate, is a coupling agent used in the synthesis of ureas, carbamates, and carbonates. It is also used as a building block in solid-phase peptide synthesis (SPPS) to obtain altered side chain functionalities in peptides.

Application

4-Nitrophenyl chloroformate is used as:
  • A coupling agent to link the drug molecule with the polyrotaxane carrier molecule
  • As an activated ester reagent that enables the covalent coupling of a peptide onto a macromer, such as oligo(poly(ethylene glycol) fumarate) (OPF)

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Solid-phase combinatorial synthesis and cytotoxicity of 3-aryl-2, 4-quinazolindiones.
Choo Hea YP, et al.
Bioorganic & Medicinal Chemistry, 10(3), 517-523 (2002)
4-Nitrophenyl chloroformate: A versatile coupling reagent.
Sammet B, et al.
Synlett, 2009(18), 3050-3051 (2009)
Zhuojun Gu et al.
Drug development and industrial pharmacy, 41(5), 812-818 (2014-04-22)
Polyamidoamine (PAMAM) dendrimers have attracted lots of interest as drug carriers. And little study about whether pluronic-attached PAMAM dendrimers could be potential drug delivery systems has been carried on. Pluronic F127 (PF127) attached PAMAM dendrimers were designed as novel drug
Synthesis and characterization of poly (oxyethylene) modified dextrans
Hoste K, et al.
Macromolecular Rapid Communications, 15(9), 697-697 (1994)
Surface derivatization strategy for combinatorial analysis of cell response to mixtures of protein domains
C Chiang, et al.
Langmuir, 28, 548-556 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service