Skip to Content
Merck
All Photos(1)

Key Documents

909599

Sigma-Aldrich

DHSO crosslinker

Synonym(s):

3,3′-Sulfinyldi(propanehydrazide), Dihydrazide sulfoxide, Mass spectrometry-cleavable crosslinker for studying protein-protein interations

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N4O3S
CAS Number:
Molecular Weight:
222.27
UNSPSC Code:
12161502
NACRES:
NA.22

form

powder

storage temp.

2-8°C

Application

DHSO Crosslinker is a homobifunctional, acidic residue-targeting, sulfoxide-containing crosslinker for analysis of protein-protein interactions (PPIs) through crosslinking mass spectrometry (XL-MS). DHSO possesses two hydrazide-reactive groups for targeting aspartic acid (Asp) and glutamic acid (Glu) and two symmetrical C-S cleavable bonds adjacent to the central sulfoxide. While former amine-reactive reagents react with lysine (Lys) residues in proximity to the PPI, targeting acidic residues provides further opportunity to characterize protein interaction surfaces at which Asp and Glu are often prevalent. Additionally, the 12.4 Å spacer yields tagged peptides post-cleavage for unambiguous identification by collision-induced dissociation in tandem MS. DHSO Crosslinker provides complementary data to Lys-targeting reagents and will find wide utility in the elucidation of PPIs, study of proteins that function as complexes, quantification of structural dynamics, and quest for targeting "undruggable" protein targets.

Legal Information

Subject to US Patent Application #15/613,065 of the Regents of the University of California

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Craig B Gutierrez et al.
Analytical chemistry, 88(16), 8315-8322 (2016-07-16)
Cross-linking mass spectrometry (XL-MS) has become a powerful strategy for defining protein-protein interactions and elucidating architectures of large protein complexes. However, one of the inherent challenges in MS analysis of cross-linked peptides is their unambiguous identification. To facilitate this process
Craig Gutierrez et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(8), 4088-4098 (2020-02-09)
The COP9 signalosome (CSN) is an evolutionarily conserved eight-subunit (CSN1-8) protein complex that controls protein ubiquitination by deneddylating Cullin-RING E3 ligases (CRLs). The activation and function of CSN hinges on its structural dynamics, which has been challenging to decipher by
Craig B Gutierrez et al.
Analytical chemistry, 90(12), 7600-7607 (2018-05-25)
Cross-linking mass spectrometry (XL-MS) has become an emerging technology for defining protein-protein interactions (PPIs) and elucidating architectures of large protein complexes. Up to now, the most widely used cross-linking reagents target lysines. Although such reagents have been successfully applied to

Articles

Sulfoxide-containing MS-cleavable cross-linkers capture protein-protein interactions in native cellular environments, aiding PPI identification.

Sulfoxide-containing MS-cleavable cross-linkers capture protein-protein interactions in native cellular environments, aiding PPI identification.

Sulfoxide-containing MS-cleavable cross-linkers capture protein-protein interactions in native cellular environments, aiding PPI identification.

Sulfoxide-containing MS-cleavable cross-linkers capture protein-protein interactions in native cellular environments, aiding PPI identification.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service