Skip to Content
Merck
All Photos(2)

Key Documents

901342

Sigma-Aldrich

PPF

Synonym(s):

2,8-Bis(diphenylphosphineoxide)dibenzofuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H26O3P2
CAS Number:
Molecular Weight:
568.54
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

97%

Quality Level

form

powder or crystals

SMILES string

[P](=O)(c7ccccc7)(c6ccccc6)c1cc2c([o]c3c2cc(cc3)[P](=O)(c5ccccc5)c4ccccc4)cc1

InChI key

AIAJGVRFXREWPK-UHFFFAOYSA-N

General description

A dibenzofuran-based host material for blue electrophosphorescence including thermally activated delayed fluorescence (TADF) OLEDs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sae Youn Lee et al.
Dalton transactions (Cambridge, England : 2003), 44(18), 8356-8359 (2015-01-20)
Thermally activated delayed fluorescence (TADF) materials based on benzoylbenzophenone, AcPmBPX and PxPmBPX, were designed and synthesized. Organic light-emitting diodes using these materials as emitters exhibited high external electroluminescence quantum efficiencies of up to 11%.
Full-Color Delayed Fluorescence Materials Based on Wedge-Shaped Phthalonitriles and Dicyanopyrazines: Systematic Design, Tunable Photophysical Properties, and OLED Performance.
Park IS, et al.
Advances in Functional Materials, 26(11), 1813-1821 (2016)
Paul A Vecchi et al.
Organic letters, 8(19), 4211-4214 (2006-09-08)
Dibenzofuran (DBF) is converted to a vacuum-sublimable, electron-transporting host material via 2,8-substitution with diphenylphosphine oxide moieties. Close pi-pi stacking and the inductive influence of P=O moieties impart favorable electron-transport properties without lowering the triplet energy. A maximum external quantum efficiency

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service