Skip to Content
Merck
All Photos(1)

Key Documents

914819

Sigma-Aldrich

Cesium iodide

AnhydroBeads, 99.999% trace metals basis, (Perovskite grade)

Synonym(s):

Cesium monoiodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CsI
CAS Number:
Molecular Weight:
259.81
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
crystals

product line

AnhydroBeads

Quality Level

Assay

99.999% trace metals basis

form

crystals

particle size

-10 mesh

mp

626 °C (lit.)

density

4.51 g/mL at 25 °C (lit.)

SMILES string

[I-].[Cs+]

InChI

1S/Cs.HI/h;1H/q+1;/p-1

InChI key

XQPRBTXUXXVTKB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Cesium iodide anhydrous can be used as a precursor or component in the synthesis of the perovskite absorber layer in perovskite solar cells. By introducing cesium iodide into the perovskite composition, the bandgap of the material can be tuned to better match the solar spectrum, optimizing the light absorption and energy conversion efficiency of the solar cell.
Cesium iodide finds application in synthesis of perovksites based photovoltaic materials. Our perovskite grade CsI can readily be dissolved in 1:1 vol DMF/DMSO to yield 1M solution.

Features and Benefits

Frequently used in devices such as phosphor screens for medical imaging, scintillators, calorimeters and a variety of particle detectors.

Packaging

Packaged in ampules

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cesium Iodide Interface Modification for High Efficiency, High Stability and Low Hysteresis Perovskite Solar Cells
Han F, et al.
Electrochimica Acta, 236, 122-130 (2017)
Kang Wang et al.
Nature communications, 9(1), 4544-4544 (2018-11-02)
As the black cesium lead iodide (CsPbI3) tends to transit into a yellow δ-phase at ambient, it is imperative to develop a stabilized black phase for photovoltaic applications. Herein, we report a distorted black CsPbI3 film by exploiting the synergistic

Articles

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service