Skip to Content
Merck
All Photos(1)

Key Documents

900854

Sigma-Aldrich

FBR

Synonym(s):

5,5′-[(9,9-Dioctyl-9H-fluorene-2,7-diyl)bis(2,1,3-benzothiadiazole-7,4-diylmethylidyne)]bis[3-ethyl-2-thioxo-4-thiazolidinone]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H56N6O2S6
CAS Number:
Molecular Weight:
1001.44
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

description

Band gap: 1.59 eV

Quality Level

Assay

≥97%

form

solid

solubility

chlorobenzene: soluble
chloroform: soluble
dichlorobenzene: soluble

Orbital energy

HOMO -5.61 eV 
LUMO -4.02 eV 

SMILES string

S1\C(=C/c2c3n[s]nc3c(cc2)c4cc5c(cc4)c6c(cc(cc6)c7c8n[s]nc8c(cc7)\C=C9/SC(=S)N(C/9=O)CC)C5(CCCCCCCC)CCCCCCCC)\C(=O)N(C1=S)CC

InChI

1S/C53H56N6O2S6/c1-5-9-11-13-15-17-27-53(28-18-16-14-12-10-6-2)41-29-33(37-23-21-35(45-47(37)56-66-54-45)31-43-49(60)58(7-3)51(62)64-43)19-25-39(41)40-26-20-34(30-42(40)53)38-24-22-36(46-48(38)57-67-55-46)32-44-50(61)59(8-4)52(63)65-44/h19-26,29-32H,5-18,

InChI key

OLBOENQKGKQCJD-XOTMQJHLSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derya Baran et al.
Nature materials, 16(3), 363-369 (2016-11-22)
Technological deployment of organic photovoltaic modules requires improvements in device light-conversion efficiency and stability while keeping material costs low. Here we demonstrate highly efficient and stable solar cells using a ternary approach, wherein two non-fullerene acceptors are combined with both
Sarah Holliday et al.
Journal of the American Chemical Society, 137(2), 898-904 (2014-12-30)
A novel small molecule, FBR, bearing 3-ethylrhodanine flanking groups was synthesized as a nonfullerene electron acceptor for solution-processed bulk heterojunction organic photovoltaics (OPV). A straightforward synthesis route was employed, offering the potential for large scale preparation of this material. Inverted

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service