Skip to Content
Merck
All Photos(2)

Key Documents

685070

Sigma-Aldrich

F8T2

99.9%

Synonym(s):

Poly(9,9-dioctylfluorene-alt-bithiophene), Poly[(9,9-dioctylfluorenyl-2,7-diyl)-co-bithiophene], Poly[[2,2′-bithiophene]-5,5′-diyl(9,9-dioctyl-9H-fluorene-2,7-diyl)]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C37H44S2)n(C8H9)2
CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

99.9%

form

powder

mol wt

average Mn >20,000

fluorescence

λex 400 nm; λem 497 nm in chloroform (at Mn = 20,000)

semiconductor properties

P-type (mobility=5×10−3 cm2/V·s)

General description

F8T2 is a fluorenated semiconducting polymer which can be used as a hole transporting layer with mobility of 0.02cm2V-1s-1. It is highly stable in vacuum and UV based environment. Its liquid crystallinity allows it to form a self-ordered nanostructure on organic thin films.
Typically soluble in THF, Dichloromethane, or Tolune. (c = 1%, typical appearance may be clear to turbid).
Polymer is end-capped with 3,5-dimethylbenzene.

Application

F8T2 can be majorly used in the fabrication of active layers for optoelectronics and energy based devices such as organic field effect transistors(OFETs), solar cells, light emitting diodes(LEDs) and electronic gas sensors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic/inorganic F8T2/GaN light emitting heterojunction.
Wu YJ, et al.
Organic Electronics, 49(3), 64-68 (2017)
Natalie Stutzmann et al.
Science (New York, N.Y.), 299(5614), 1881-1884 (2003-03-22)
The manufacture of high-performance, conjugated polymer transistor circuits on flexible plastic substrates requires patterning techniques that are capable of defining critical features with submicrometer resolution. We used solid-state embossing to produce polymer field-effect transistors with submicrometer critical features in planar
Accelerating gas adsorption on 3D percolating carbon nanotubes.
Li H, et al.
Scientific Reports, 6(13), 21313-21313 (2016)
Preparation of active layer of solar cells device by F8T2 blending with PCBM
CPMT Symposium Japan, 2010 IEEE, 3(13), 1-4 (2010)
Structural ordering in F8T2 polyfluorene thin film transistors.
Organic Field Effect Transistors II, 5217, 35-43 (2003)

Articles

The development of high-performance conjugated organic molecules and polymers has received widespread attention in industrial and academic research.

Organic photovoltaics (OPVs) represent a low-cost, lightweight, and scalable alternative to conventional solar cells. While significant progress has been made in the development of conventional bulk heterojunction cells, new approaches are required to achieve the performance and stability necessary to enable commercially successful OPVs.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service