Skip to Content
Merck
All Photos(1)

Key Documents

900064

Sigma-Aldrich

Iron oxide (II,III), nanoparticles

25 nm avg. part. size (TEM), 5 mg/mL in toluene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352303
NACRES:
NA.23
Pricing and availability is not currently available.

form

dispersion
nanoparticles

Quality Level

concentration

5 mg/mL in toluene

avg. part. size

25 nm (TEM)

functional group

oleic acid

Looking for similar products? Visit Product Comparison Guide

Application

Iron oxide nanoparticles exhibit biocompatibility and non-toxicity and are used as magnetic resonance imaging contrast agents, gene carriers for gene therapy, therapeutic agents for hyperthermia-based cancer treatment, magnetic sensing probes for in vivo diagnostics and for target specific drug delivery.[1][2][3]

Legal Information

Product of Ocean Nanotech, LLC.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F

Flash Point(C)

5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianglong Hu et al.
Dalton transactions (Cambridge, England : 2003), 44(9), 3904-3922 (2015-01-13)
Responsive polymeric assemblies and hybrid superstructures fabricated from stimuli-sensitive polymers and inorganic nanoparticles (NPs) have been the subject of extensive investigations during the past few decades due to their distinct advantages such as an improved water solubility, stimuli-responsiveness, excellent biocompatibility
Joan Estelrich et al.
International journal of molecular sciences, 16(4), 8070-8101 (2015-04-14)
In this review, we discuss the recent advances in and problems with the use of magnetically-guided and magnetically-responsive nanoparticles in drug delivery and magnetofection. In magnetically-guided nanoparticles, a constant external magnetic field is used to transport magnetic nanoparticles loaded with
Jin Xie et al.
Theranostics, 2, 122-124 (2012-01-31)
This theme issue provides a timely collection of studies on magnetic nanoparticle-based imaging, bio-sensing, therapy and/or their combinations.

Articles

Nanoparticle imaging agents enable stem cell tracking via various in vivo imaging techniques.

Nanoparticle imaging agents enable stem cell tracking via various in vivo imaging techniques.

Nanoparticle imaging agents enable stem cell tracking via various in vivo imaging techniques.

Nanoparticle imaging agents enable stem cell tracking via various in vivo imaging techniques.

See All

Questions

  1. Is the surface of the nanoparticles hydrophobic or hydrophilic?

    1 answer
    1. This product is supplied as a dispersion in toluene. The surface ligand is oleic acid which is described as primarily hydrophobic, as this ligand is a monounsaturated omega-9 fatty acid.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service