Skip to Content
Merck
All Photos(1)

Key Documents

86480

Sigma-Aldrich

Terpineol

mixture of isomers

Synonym(s):

Alpha-Terpineol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
60.0-85.0% (GC)
bp:
213-218 °C (lit.)
Pricing and availability is not currently available.

Quality Level

Assay

60.0-85.0% (GC)

form

viscous liquid

composition

α-terpineol, 60.0-85.0%
β-terpineol, 0.5-13.0%
γ-terpineol, 10.0-25.0%

impurities

≤1.0% water

refractive index

n20/D 1.484

bp

213-218 °C (lit.)

density

0.934 g/mL at 20 °C (lit.)

functional group

hydroxyl

SMILES string

CC(=C)C1CCC(C)(O)CC1.CC2=CCC(CC2)C(C)(C)O

InChI

1S/2C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11;1-8(2)9-4-6-10(3,11)7-5-9/h4,9,11H,5-7H2,1-3H3;9,11H,1,4-7H2,2-3H3

InChI key

MGABGZGUMNDCSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Terpineol has been used as a high-viscosity solvent along with ethyl cellulose as a resin to prepare dispersion paste of titanium dioxide (TiO2) that is used in dye-sensitized solar cell (DSSC) technology.[1] It can also be used in the preparation of graphene/terpineol dispersion for use in inkjet printing.[2]

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Caution

may crystallize out under cold conditions

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The optimisation of dye sensitised solar cell working electrodes for graphene and SWCNTs containing quasi-solid state electrolytes.
Byrne O, et. al.
Solar Energy, 110, 239-246 (2014)
Sheng-Chang Wang et al.
Sensors (Basel, Switzerland), 15(6), 14286-14297 (2015-06-20)
Highly sensitive H2 gas sensors were prepared using pure and Pt-loaded SnO2 nanoparticles. Thick film sensors (~35 μm) were fabricated that showed a highly porous interconnected structure made of high density small grained nanoparticles. Using Pt as catalyst improved sensor
Efficient inkjet printing of graphene.
Li J
Advanced Materials, 25(29), 3985-3992 (2013)
Khaled Sebei et al.
Biological research, 48, 7-7 (2015-02-06)
In this paper, we have studied the essential oils chemical composition of the leaves of seven Eucalyptus species developed in Tunisia. Eucalyptus leaves were picked from trees growing in different arboretums in Tunisia. Choucha and Mrifeg arboretums located in Sedjnene
Yi-Fei Sun et al.
Nanoscale, 7(25), 11173-11181 (2015-06-11)
To date, most investigations of Ni-Fe bimetallic catalysts for solid oxide fuel cells (SOFCs) have focused on materials with micro-scale particle sizes, which severely restrict their catalytic activity. In this study, we fabricated a Ni- and/or Fe-doped A-site-deficient LaSrCrO3 perovskite

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service