Skip to Content
Merck
All Photos(1)

Documents

81801

Sigma-Aldrich

Propylphosphonic anhydride solution

technical, ~50% in DMF

Synonym(s):

1-Propanephosphonic anhydride solution, 2,4,6-Tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide solution, PPACA, T3P®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H21O6P3
CAS Number:
Molecular Weight:
318.18
Beilstein:
5079255
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

form

solution

concentration

~50% in DMF

SMILES string

CCCP1(=O)OP(=O)(CCC)OP(=O)(CCC)O1

InChI

1S/C9H21O6P3/c1-4-7-16(10)13-17(11,8-5-2)15-18(12,14-16)9-6-3/h4-9H2,1-3H3

InChI key

PAQZWJGSJMLPMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propylphosphonic anhydride is a highly reactive cyclic anhydride that is commonly used as a coupling agent and water scavenger.

Application

Coupling reagent for peptide synthesis; Preparation of other amides; Synthesis of esters (depsipeptides)

Features and Benefits

Some of its unique features include:
  • Broad functional group tolerance
  • Low epimerization tendency
  • By-products are water-soluble
  • High yield and purity


Legal Information

T3P is a registered trademark of Archimica GmbH

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

143.6 °F

Flash Point(C)

62 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Kessler et al.
Liebigs Ann. Chem., 1-1 (1986)
H. Wissmann
Phosph. Sulfur Relat. Elem., 30, 645-645 (1986)
H. Wissmann et al.
Angewandte Chemie (International Edition in English), 92, 129-129 (1980)
Propylphosphonic anhydride (T3P?): an efficient reagent for the one-pot synthesis of 1, 2, 4-oxadiazoles, 1, 3, 4-oxadiazoles, and 1, 3, 4-thiadiazoles.
Augustine JK, et al.
Tetrahedron, 65(48), 9989-9996 (2009)
M. Feigel
Journal of the American Chemical Society, 108, 181-181 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service