Skip to Content
Merck
All Photos(7)

Key Documents

806625

Sigma-Aldrich

Graphene nanoplatelets

powder, polycarboxylate functionalized, hydrophilic

Synonym(s):

GNPs, graphenit-HYDRO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
UNSPSC Code:
12352103
NACRES:
NA.23

description

Dispersibility: water (high stability in aqueous medium)

Quality Level

form

powder

composition

Carbon, >70 wt. %
Oxygen, >10 wt. %

Looking for similar products? Visit Product Comparison Guide

General description

Graphene nanoplatelets are “pure carbon” very lipophilic, with a high tendency to aggregate and poor interactions with the surrounding matrix, hence difficult to disperse in processes and supports such as epoxy resin, thermoplastics or solvents such as water. The chemical modification on the surface of graphite nanoplatelet open new properties allowing good interaction with the targeted matrix. Without these chemical modifications the dispersion step became a difficult task and quite often do not produce the expected benefits.

Application

  • Graphene (nano)composite materials
  • Conductive coatings
  • Conductive Inks
  • Energy Storage
  • Electrode materials

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-state functionalization of graphene with amino acids toward water-dispersity: implications on a composite with polyaniline and its characteristics as a supercapacitor electrode material
Erdenedelger et al.
Journal of Material Chemistry A, 2(31), 12526-12534 (2014)
Maria Suarez-Diez et al.
Scientific reports, 10(1), 3232-3232 (2020-02-26)
Graphene nanomaterials have attracted a great interest during the last years for different applications, but their possible impact on different biological systems remains unclear. Here, an assessment to understand the toxicity of commercial polycarboxylate functionalized graphene nanoplatelets (GN) on the
Edge-carboxylated graphene nanoplatelets as oxygen-rich metal-free cathodes for organic dye-sensitized solar cells
Ju et al.
Energy & Environmental Science, 7(3), 1044-1052 (2014)
Chang-Hsiao Chen et al.
Nanomedicine : nanotechnology, biology, and medicine, 9(5), 600-604 (2013-01-26)
A graphene-based flexible microprobe developed by microelectromechanical system technology shows high resolution for the detection of electrophysiological signals from various bio-objects. The hydrophilization post-treatment using steam plasma was performed on the graphene surface to decrease the interfacial impedance between graphene

Articles

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service