Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Graphene nanoplatelets
Empirical Formula (Hill Notation):
C
CAS Number:
7782-42-5
Molecular Weight:
12.01
EC Number:
231-955-3
Products
Brand
Sigma-Aldrich (6)
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Particle Size (µm)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
900407
Graphene nanoplatelets,
<2 μm particle size
Expand
View Pricing
900413
Graphene nanoplatelets,
25 μm particle size
Expand
View Pricing
900439
Graphene nanoplatelets,
<2 μm particle size
Expand
View Pricing
900412
Graphene nanoplatelets,
5 μm particle size
Expand
View Pricing
900411
Graphene nanoplatelets,
25 μm particle size
Expand
View Pricing
900410
Graphene nanoplatelets,
15 μm particle size
Expand
View Pricing