Skip to Content
Merck
All Photos(2)

Key Documents

791504

Sigma-Aldrich

Carbon nanotube, single-walled

greener alternative

conductive aqueous ink, 0.9-1.1 mg/mL SWCNT(concentration by Absorbance at 854 nm)

Synonym(s):

CG300-Aqueous Ink, SWCNT Ink, SWNT Ink, SWeNT AC200, Single-Walled Carbon Nanotube Ink

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

product name

Carbon nanotube, single-walled, conductive aqueous ink, 0.9-1.1 g/L (SWCNT concentration by Absorbance at 854 nm), avg. no. of layers, 1

form

dispersion in H2O (black liquid)

feature

avg. no. of layers 1

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

0.9-1.1 g/L (SWCNT concentration by Absorbance at 854 nm)

sheet resistance

<600 Ω/sq, at 85% VLT (ohm/square) (by 4-point probe on prepared film by rod coating)

viscosity

3.0 mPa.s (at 10 sec-1 shear rate)

density

1 g/cm3

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Click here for more information.

Application

This conductive ink is aqueous based and contains highly conductive single wall carbon nanotubes and is formulated for spray coating.This ink is primarily intended for making highly transparent conductive films on a variety of substrates.

Legal Information

Product of Chasm Advanced Materials, Inc.
Signis is a registered trademark of Chasm Advanced Materials, Inc.
CoMoCAT is a trademark of Chasm Advanced Materials, Inc.
CHASM is a trademark of Chasm Advanced Materials, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Functional materials for printed electronics applications enable flexible displays, RFID tags, and biomedical sensors.

Functional materials for printed electronics applications enable flexible displays, RFID tags, and biomedical sensors.

Functional materials for printed electronics applications enable flexible displays, RFID tags, and biomedical sensors.

Functional materials for printed electronics applications enable flexible displays, RFID tags, and biomedical sensors.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service