776270
n-Propylmagnesium chloride solution
1.0 M in 2-methyltetrahydrofuran
Synonym(s):
Propylmagnesium chloride solution
About This Item
Recommended Products
form
solution
reaction suitability
reaction type: Grignard Reaction
greener alternative product characteristics
Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
concentration
1.0 M in 2-methyltetrahydrofuran
density
0.896 g/mL at 25 °C
greener alternative category
SMILES string
CCC[Mg]Cl
InChI
1S/C3H7.ClH.Mg/c1-3-2;;/h1,3H2,2H3;1H;/q;;+1/p-1
InChI key
AMXVKMXLDJXAPM-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
2-Methyltetrahydrofuran (2-MeTHF): A Biomass-Derived Solvent with Broad Application in Organic Chemistry
Packaging
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Water-react 2
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 2
Flash Point(F)
-22.0 °F - closed cup
Flash Point(C)
-30 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service