Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbon nanotube, single-walled
Synonym(s):
SWCNT, SWNT, Single wall carbon nanotube
CAS Number:
308068-56-6
Products
Brand
Sigma-Aldrich (11)
Color
Manufacturer
Melting Point (°C)
Physical Form
Purity
Quality Segment
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
773735
Carbon nanotube, single-walled,
(6,5) chirality, ≥95% carbon basis (≥95% as carbon nanotubes), 0.78 nm average diameter
Expand
View Pricing
900711
Carbon nanotube, single-walled,
≥98% carbon basis, avg. no. of layers, 1
Expand
View Pricing
775533
Carbon nanotube, single-walled,
≥95% carbon basis (≥99% as carbon nanotubes), 0.84 nm average diameter, avg. no. of layers, 1
Expand
View Pricing
805033
Carbon nanotube, single-walled,
carbon ≥80 %, diam. 1.2-2.0 nm, avg. no. of layers, 1
Expand
View Pricing
724777
Carbon nanotube, single-walled,
≥90% carbon basis (≥80% as carbon nanotubes), 1-2 nm diameter, avg. no. of layers, 1
Expand
View Pricing
750522
Carbon nanotube, single-walled,
98% (Semiconducting), avg. no. of layers, 1
Expand
View Pricing
704121
Carbon nanotube, single-walled,
(7,6) chirality, ≥90% carbon basis (≥77% as carbon nanotubes), 0.83 nm average diameter, avg. no. of layers, 1
Expand
View Pricing
901634
Carbon nanotube, single-walled,
Low metal content, avg. no. of layers, 1
Expand
View Pricing
750530
Carbon nanotube, single-walled,
98% (Metallic), avg. no. of layers, 1
Expand
View Pricing
755710
Carbon nanotube, single-walled,
>70% (TGA), avg. no. of layers, 1
Expand
View Pricing
750492
Carbon nanotube, single-walled,
<3.5% Metal Catalyst, avg. no. of layers, 1
Expand
View Pricing