Skip to Content
Merck
All Photos(1)

Documents

773174

Sigma-Aldrich

Fmoc-N-Me-Tyr(tBu)-OH

97%

Synonym(s):

fmoc-N-me-tyr(tbu)-oh

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H31NO5
CAS Number:
Molecular Weight:
473.56
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

97%

form

powder or crystals

optical activity

[α]22/D -49.0°, c = 0.5% in DMF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

186-191 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CN([C@@H](Cc1ccc(OC(C)(C)C)cc1)C(O)=O)C(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C29H31NO5/c1-29(2,3)35-20-15-13-19(14-16-20)17-26(27(31)32)30(4)28(33)34-18-25-23-11-7-5-9-21(23)22-10-6-8-12-24(22)25/h5-16,25-26H,17-18H2,1-4H3,(H,31,32)/t26-/m0/s1

InChI key

WTLSDEYZKFJXFT-SANMLTNESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service