Skip to Content
Merck

8.52289

Fmoc-N-Me-Ser(tBu)-OH

Novabiochem®

Synonym(s):

Fmoc-N-Me-Ser(tBu)-OH, Fmoc-N-α-methyl-O-t-butyl-L-serine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H27NO5
CAS Number:
Molecular Weight:
397.46
UNSPSC Code:
12352209
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥95.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

N([C@@H](COC(C)(C)C)C(=O)O)(C)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C23H27NO5/c1-23(2,3)29-14-20(21(25)26)24(4)22(27)28-13-19-17-11-7-5-9-15(17)16-10-6-8-12-18(16)19/h5-12,19-20H,13-14H2,1-4H3,(H,25,26)/t20-/m0/s1

InChI key

PQSAXALAXPNFMG-FQEVSTJZSA-N

General description

Building block for introduction of N-α-methyl-serine amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 98.5 % (a/a)
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library