769533
Chromium(III) oxide
ultra dry
Synonym(s):
Chromia
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Cr2O3
CAS Number:
Molecular Weight:
151.99
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
powder
Quality Level
quality
ultra dry
cation traces
Al: ≤50 ppm
Ba: ≤30 ppm
Cu: ≤10 ppm
K: ≤100 ppm
Mg: ≤50 ppm
Na: ≤500 ppm
Pb: ≤20 ppm
SMILES string
O=[Cr]O[Cr]=O
InChI
1S/2Cr.3O
InChI key
QDOXWKRWXJOMAK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Chromium oxide finds application as green pigment material. It is also used as electrode material in lithium ion batteries and as precursor for synthesis of functional ceramics and oxides.
Used in a solid-state reaction to form the new ternary oxide, CrVMoO7, can be used in selective oxidation.
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Investigation of Cr2O3 as Anode Materials for Lithium-Ion Batteries by Electrochemical Impedance Spectroscopy
Zhao, X., et al.
Journal of the Electrochemical Society, 162(7), A1156-A1162 (2015)
Magnetic and microstructural properties of LiCrO2.Cr2O3 system by doping of the boron ions
Demirel, S., et al.
J. Mater. Sci., 26(11), 9178-9184 (2015)
Preparation and color performance control of Cr2O3 green pigment through thermal decomposition of chromium hydroxide precursor
Zhang, HL., et al.
Materials Letters, 117, 244-247 (2014)
Effects on dielectric and magnetic properties of Cr-doped Bi0.9Ba0.1Fe1-xCrxO3 ceramic
Li, Q., et al.
Journal of Materials Science: Materials in Electronics, 26(11), 8887-8891 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service