Skip to Content
Merck
All Photos(1)

Key Documents

76420

Sigma-Aldrich

1-Butyl-3-methylimidazolium trifluoromethanesulfonate

≥95.0% (H-NMR)

Synonym(s):

BMIM Otf

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15F3N2O3S
CAS Number:
Molecular Weight:
288.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (H-NMR)

form

liquid

refractive index

n20/D 1.434

density

1.292 g/mL at 20 °C (lit.)

functional group

fluoro
triflate

SMILES string

[O-]S(=O)(=O)C(F)(F)F.CCCCn1cc[n+](C)c1

InChI

1S/C8H15N2.CHF3O3S/c1-3-4-5-10-7-6-9(2)8-10;2-1(3,4)8(5,6)7/h6-8H,3-5H2,1-2H3;(H,5,6,7)/q+1;/p-1

InChI key

FRZPYEHDSAQGAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

1-Butyl-3-methylimidazolium trifluoromethanesulfonate is an aprotic neutral ionic liquid.

Application

1-Butyl-3-methylimidazolium trifluoromethanesulfonate can be used as an alternative to lithium perchlorate-diethyl ether mixture in the Diels-Alder reaction.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deep fuels desulfurization and denitrogenation using 1-butyl-3-methylimidazolium trifluoromethanesulfonate
Ke?dra-Krolik, Karolina, et al.
Energy and Fuels, 25.4, 1559-1565 (2011)
Activity coefficients at infinite dilution measurements for organic solutes and water in the ionic liquid 1-butyl-3-methylimidazolium trifluoromethanesulfonate
Doman?ska, Urszula, and Andrzej Marciniak
The Journal of Physical Chemistry B, 112.35, 11100-11105 (2008)
Activity coefficients at infinite dilution of alkanes, alkenes, and alkyl benzenes in 1-butyl-3-methylimidazolium trifluoromethanesulfonate using gas? liquid chromatography
Ge, Ming-Lan, et al.
Journal of Chemical and Engineering Data, 52.6, 2257-2260 (2007)
tert-Amyl ethyl ether separation from its mixtures with ethanol using the 1-butyl-3-methylimidazolium trifluoromethanesulfonate ionic liquid: Liquid? liquid equilibrium
Arce, Alberto, Oscar Rodriguez, and Ana Soto
Industrial & Engineering Chemistry Research, 43.26, 8323-8327 (2004)
Ion-conductive polymer membranes containing 1-butyl-3-methylimidazolium trifluoromethanesulfonate and 1-ethylimidazolium trifluoromethanesulfonate.
Schauer J, et al.
Journal of Membrane Science, 367(1), 332-339 (2011)

Articles

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service