Skip to Content
Merck
1-Butyl-3-methylimidazolium trifluoromethanesulfonate ≥95.0% (H-NMR)

1-Butyl-3-methylimidazolium trifluoromethanesulfonate

1-Butyl-3-methylimidazolium trifluoromethanesulfonate ≥95.0% (H-NMR)
Synonym(s):
BMIM Otf
Empirical Formula (Hill Notation):
C9H15F3N2O3S
CAS Number:
Molecular Weight:
288.29

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade