Skip to Content
Merck
All Photos(3)

Key Documents

760676

Sigma-Aldrich

Dibenzocyclooctyne-PEG4-maleimide

for Copper-free Click Chemistry

Synonym(s):

Polyethylene glycol, DBCO-PEG4-maleimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H42N4O9
Molecular Weight:
674.74
MDL number:
UNSPSC Code:
12161502
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

solid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

functional group

maleimide

storage temp.

−20°C

SMILES string

O=C(CCNC(CCOCCOCCOCCOCCNC(CCN1C(C=CC1=O)=O)=O)=O)N2CC3=C(C=CC=C3)C#CC4=C2C=CC=C4

InChI

1S/C36H42N4O9/c41-32(14-18-39-34(43)11-12-35(39)44)38-17-20-47-22-24-49-26-25-48-23-21-46-19-15-33(42)37-16-13-36(45)40-27-30-7-2-1-5-28(30)9-10-29-6-3-4-8-31(29)40/h1-8,11-12H,13-27H2,(H,37,42)(H,38,41)

InChI key

VVFZXPZWVJMYPX-UHFFFAOYSA-N

Application

Maleimide functionalized cyclooctyne derivative for incorporation of the cyclooctyne moiety into thiol containing compounds or biomolecules. Cyclooctynes are useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. This dibenzocyclooctyne will react with azide functionalized compounds or biomolecules without the need for a Cu(I) catalyst to result in a stable triazole linkage. The PEG lipophilic spacer helps to reduce aggregation and precipitation problems when labeling biomolecules.
Applications Include:
  • Protein-peptide conjugates
  • Antibody-enzyme or antibody-drug conjugates
  • Protein or peptide-oligonucleotide conjugates
  • Surface modification

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nano-scale alignment of proteins on a flexible DNA backbone.
Nojima T, et al.
PLoS ONE, 7(12), e52534-e52534 (2012)
Ning, X., et al.
Angewandte Chemie (International Edition in English), 47, 2253-2255 null
Efficient cargo delivery into adult brain tissue using short cell-penetrating peptides
Kizil C, et al.
PLoS ONE, 10(4), e0124073-e0124073 (2015)
Xiaowen Liu et al.
Biomacromolecules, 17(1), 173-182 (2015-12-15)
Intracellularly-acting therapeutic proteins are considered promising alternatives for the treatment of various diseases. Major limitations of their application are low efficiency of intracellular delivery and possible reduction of protein activity during derivatization. Herein, we report pH-sensitive covalent modification of proteins
Heather D Kamp et al.
NPJ vaccines, 5, 33-33 (2020-05-08)
A growing global health concern, Lyme disease has become the most common tick-borne disease in the United States and Europe. Caused by the bacterial spirochete Borrelia burgdorferi sensu lato (sl), this disease can be debilitating if not treated promptly. Because

Articles

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service