Skip to Content
Merck
All Photos(1)

Documents

760706

Sigma-Aldrich

Sulfo-dibenzocyclooctyne-biotin conjugate

for Copper-free Click Chemistry

Synonym(s):

Sulfo-DBCO-biotin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H50N6O7S2
Molecular Weight:
754.96
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

storage temp.

−20°C

SMILES string

CC[NH+](CC)CC.[O-]S(=O)(=O)C(CNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)C(=O)NCCC(=O)N3Cc4ccccc4C#Cc5ccccc35

InChI

1S/C31H35N5O7S2.C6H15N/c37-27(12-6-5-11-25-29-23(19-44-25)34-31(40)35-29)33-17-26(45(41,42)43)30(39)32-16-15-28(38)36-18-22-9-2-1-7-20(22)13-14-21-8-3-4-10-24(21)36;1-4-7(5-2)6-3/h1-4,7-10,23,25-26,29H,5-6,11-12,15-19H2,(H,32,39)(H,33,37)(H2,34,35,40)(H,41,42,43);4-6H2,1-3H3/t23-,25-,26?,29-;/m0./s1

InChI key

RACLEVCKGUBCTH-GESOYTMLSA-N

General description

This sulfonated azadibenzocyclooctyne-biotin derivative is a highly water soluble and versatile biotinylation reagent for labeling azide containing molecules or compounds. Cyclooctynes are useful in strain-promoted copper-free azide-alkyne cycloaddition reactions.This dibenzocyclooctyne will react with azide-functionalized compounds or biomolecules without the need for a Cu(I) catalyst to result in a stable triazole linkage. Subsequent labeled molecules can be purified using streptavidin or avidin affinity reagents.

Application

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

Features and Benefits

More efficient labeling will occur at higher temperatures and with greater excess of biotin reagent. Typical reaction times are up to 4 hours, but longer incubation periods should be used if concentration is low or if low labeling is observed. Product is most efficient when first dissolved in a water miscible organic solvent and then diluted in aqueous reaction buffer when needed. Stock solutions in DMSO have been shown to be stable in the freezer for up to 3 months.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning, X., et al.
Angewandte Chemie (International Edition in English), 47, 2253-2255 null
Guillaume Charron et al.
Journal of the American Chemical Society, 131(13), 4967-4975 (2009-03-14)
Fatty-acylation of proteins in eukaryotes is associated with many fundamental cellular processes but has been challenging to study due to limited tools for rapid and robust detection of protein fatty-acylation in cells. The development of azido-fatty acids enabled the nonradioactive
Andrew Vila et al.
Chemical research in toxicology, 21(2), 432-444 (2008-02-01)
Polyunsaturated fatty acids (PUFA) are primary targets of free radical damage during oxidative stress. Diffusible electrophilic alpha,beta-unsaturated aldehydes, such as 4-hydroxynonenal (HNE), have been shown to modify proteins that mediate cell signaling (e.g., IKK and Keap1) and alter gene expression
Daniela C Dieterich et al.
Nature protocols, 2(3), 532-540 (2007-04-05)
A major aim of proteomics is the identification of proteins in a given proteome at a given metabolic state. This protocol describes the step-by-step labeling, purification and detection of newly synthesized proteins in mammalian cells using the non-canonical amino acid
Nicholas J Agard et al.
ACS chemical biology, 1(10), 644-648 (2006-12-19)
Detection of metabolites and post-translational modifications can be achieved using the azide as a bioorthogonal chemical reporter. Once introduced into target biomolecules, either metabolically or through chemical modification, the azide can be tagged with probes using one of three highly

Articles

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service