Skip to Content
Merck
All Photos(1)

Key Documents

09719

Sigma-Aldrich

Ammonium iron(II) sulfate hexahydrate

BioUltra, ≥99.0% (RT)

Synonym(s):

Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate, Mohr’s salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2Fe(SO4)2 · 6H2O
CAS Number:
Molecular Weight:
392.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
grade:
reagent grade
solubility:
H2O: 0.5 M at 20 °C, clear, faintly yellow-green

grade

reagent grade

Quality Level

product line

BioUltra

Assay

≥99.0% (RT)

impurities

insoluble matter, passes filter test

pH

2.5-4.0 (25 °C, 0.5 M in H2O)

mp

100 °C (dec.) (lit.)

solubility

H2O: 0.5 M at 20 °C, clear, faintly yellow-green

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤20 mg/kg

cation traces

Cd: ≤5 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤5 mg/kg
Fe3+: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤100 mg/kg
Mn: ≤500 mg/kg
Na: ≤50 mg/kg
Ni: ≤100 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

N.N.O.O.O.O.O.O.[Fe++].OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Fe.2H3N.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h;2*1H3;2*(H2,1,2,3,4);6*1H2/q+2;;;;;;;;;;/p-2

InChI key

MQLVWQSVRZVNIP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium iron(II) sulfate hexahydrate has been used in a study to assess the catalytic effect of different forms of iron in purification of single-walled carbon nanotubes. Ammonium iron(II) sulfate hexahydrate has also been used in a study to determine trace amounts of Se(IV) in seawater.
In culture media for the detection of sulfate-reducing bacteria;

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoko Suzuki et al.
Journal of nanoscience and nanotechnology, 10(6), 3924-3928 (2010-04-02)
In the arc plasma jet (APJ) method, a large amount of soot including single wall carbon nanotubes (SWNTs) can be produced in a short time. However, as-grown soot contains a lot of impurities, such as metallic particles used as catalyst
Gong Zhengjun et al.
Talanta, 66(4), 1012-1017 (2008-10-31)
A simple, accurate, sensitive and selective flow injection catalytic kinetic spectrophotometric method for rapid determination of trace amounts of selenium is proposed in this paper. The proposed method is based on the accelerating effect of Se(IV) on the reaction of
Yu-Jung Chang et al.
Nutrition (Burbank, Los Angeles County, Calif.), 21(4), 520-524 (2005-04-07)
This study examined whether iron accumulated in ferritin-producing recombinant microbes is bioavailable to rats with iron deficiency. Rats induced with iron deficiency were treated with iron preparations of ferrous ammonium sulfate, horse spleen ferritin, control yeast, and ferritin-producing recombinant yeast
Ubon Cha'on et al.
Toxicological sciences : an official journal of the Society of Toxicology, 96(1), 194-201 (2006-12-19)
The aim of this study is to identify the biochemical mechanism of phosphine toxicity and resistance, using Caenorhabditis elegans as a model organism. To date, the precise mode of phosphine action is unclear. In this report, we demonstrate the following
Kazumasa Kumamoto et al.
Free radical research, 39(9), 987-993 (2005-08-10)
Identification of free radicals was performed for the reaction mixtures of autoxidized 1,2-dilinoleoylphosphatidylcholine (DLPC) with ferrous ions (or DLPC hydroperoxide with ferrous ions) and of DLPC with soybean lipoxygenase using electron spin resonance (ESR), high performance liquid chromatography (HPLC)--ESR and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service