Skip to Content
Merck
All Photos(1)

Documents

747939

Sigma-Aldrich

Indium tin oxide

≥99.5% trace metals basis, 18 nm particle size (SEM), 20 wt. % in H2O

Synonym(s):

ITO

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.5% trace metals basis

form

dispersion

composition

In2O3, 90%
SnO2, 10%

concentration

20 wt. % in H2O

particle size

18 nm (SEM)

mp

1910 °C (lit.)

density

1.1682 g/mL at 25 °C
1.2 g/mL at 25 °C (lit.)

SMILES string

O=[SnH2].O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Indium Tin oxide nanoparticles have been investigated for use in:
  • transparent electrodes
  • the formation of conductive paper and organic contaminants
  • the formation of transparent conductive oxide films and magnetic nanocomposites
  • electrochromic materials and in biomedical applications

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conductive paper fabricated by layer-by-layer assembly of polyelectrolytes and ITO nanoparticles.
Peng C, et al.
Nanotechnology, 19(50) (2008)
Characterization of dip-coated ITO films derived from nanoparticles synthesized by?low-pressure spray pyrolysis.
Ogi T, et al.
Journal of Nanoparticle Research, 8(3-4), 343-350 (2006)
Progress in Electrochromics and Thermochromics: Two New Applications Involving ITO Nanoparticles.
Li S, et al.
Society of Vacuum Coaters; 55th Annual Technical Conference Proceedings, 41-46 (2012)
Generation of transparent conductive electrodes by laser consolidation of LIFT printed ITO nanoparticle layers.
Baum M, et al.
Applied Physics. A, Materials Science & Processing, 111(3), 799-805 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service