Skip to Content
Merck
All Photos(2)

Documents

473677

Sigma-Aldrich

(Aminomethyl)polystyrene

200-400 mesh, extent of labeling: 4.0 mmol/g loading, 2 % cross-linked

Synonym(s):

AM-polystyrene, Benzylamine polymer-bound, Poly(styrene-co-divinylbenzene), aminomethylated, Poly(styrene-co-vinylbenzylamine-co-divinylbenzene, Polystyrene crosslinked with divinylbenzene, aminomethylated

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
13102026
PubChem Substance ID:
NACRES:
NA.22

crosslinking

2 % cross-linked

reaction suitability

reaction type: Boc solid-phase peptide synthesis

extent of labeling

4.0 mmol/g loading

particle size

200-400 mesh

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Scavenges acids, acid chlorides, anhydrides, aldehydes and other electrophiles

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service