Skip to Content
Merck
All Photos(2)

Key Documents

747319

Sigma-Aldrich

Iron oxide(II,III), magnetic nanoparticles solution

10 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL Fe in H2O, dispersion

Synonym(s):

Fe NP PEG, FexOy, Magnetic iron oxide nanocrystals, Magnetite, Superparamagnetic iron oxide nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Fe3O4
CAS Number:
Molecular Weight:
231.53
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

dispersion
nanoparticles

concentration

1 mg/mL Fe in H2O

magnetization

>45 emu/g, at room temperature; under 4500 Oe

color

brown

avg. part. size

10 nm (TEM)

density

0.996 g/mL at 25 °C
~1 g/mL at 25 °C

functional group

PEG

storage temp.

2-8°C

SMILES string

O=[Fe].O=[Fe]O[Fe]=O

InChI

1S/3Fe.4O

InChI key

SZVJSHCCFOBDDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Concentration 5mg/ml includes total weight nanocrystals plus ligands.

Application

These magnetic nanoparticles are typically used in imaging or as contrast agents. The surface functionality allows for different ligation or further functionalization.

Do not freeze.

Legal Information

Product of Ocean Nanotech; LLC.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Lohße et al.
Journal of bacteriology, 196(14), 2658-2669 (2014-05-13)
Biosynthesis of bacterial magnetosomes, which are intracellular membrane-enclosed, nanosized magnetic crystals, is controlled by a set of >30 specific genes. In Magnetospirillum gryphiswaldense, these are clustered mostly within a large conserved genomic magnetosome island (MAI) comprising the mms6, mamGFDC, mamAB
Aude Picard et al.
Nature communications, 6, 6277-6277 (2015-02-19)
Twisted stalks are organo-mineral structures produced by some microaerophilic Fe(II)-oxidizing bacteria at O2 concentrations as low as 3 μM. The presence of these structures in rocks having experienced a diagenetic history could indicate microbial Fe(II)-oxidizing activity as well as localized
Yingjie Li et al.
Journal of bacteriology, 196(14), 2552-2562 (2014-05-06)
The biomineralization of magnetosomes in Magnetospirillum gryphiswaldense and other magnetotactic bacteria occurs only under suboxic conditions. However, the mechanism of oxygen regulation and redox control of biosynthesis of the mixed-valence iron oxide magnetite [FeII(FeIII)2O4] is still unclear. Here, we set
Tianyuzi Li et al.
International journal of nanomedicine, 10, 3779-3790 (2015-06-18)
Regimen adherence, systemic toxicities, and limited drug penetrance to viral reservoirs are obstacles limiting the effectiveness of antiretroviral therapy (ART). Our laboratory's development of the monocyte-macrophage-targeted long-acting nanoformulated ART (nanoART) carriage provides a novel opportunity to simplify drug-dosing regimens. Progress
Faheem Muhammad et al.
Colloids and surfaces. B, Biointerfaces, 123, 506-514 (2014-12-03)
Simultaneous delivery of multiple therapeutic agents is of great importance for effective chemotherapy due of its well-known drug synergism and suppression to chemoresistance. We report a new theranostic nanoformulation to shuttle multiple chemotherapeutic agents for successfully exterminating cancer cells. This

Articles

Iron oxide nanoparticles find diverse applications in magnetic data storage, biosensing, and drug delivery due to their properties.

Iron oxide nanoparticles find diverse applications in magnetic data storage, biosensing, and drug delivery due to their properties.

Iron oxide nanoparticles find diverse applications in magnetic data storage, biosensing, and drug delivery due to their properties.

Iron oxide nanoparticles find diverse applications in magnetic data storage, biosensing, and drug delivery due to their properties.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service