Skip to Content
Merck
All Photos(1)

Key Documents

725331

Sigma-Aldrich

Iron oxide(II,III), magnetic nanoparticles solution

5 nm avg. part. size, 5 mg/mL in H2O

Synonym(s):

Magnetic iron oxide nanocrystals, Magnetite, Superparamagnetic iron oxide nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Fe3O4
CAS Number:
Molecular Weight:
231.53
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

dispersion
nanoparticles

Quality Level

concentration

5 mg/mL in H2O

magnetization

>25 emu/g, at 4500Oe

particle size

4-6 nm (TEM)

avg. part. size

5 nm

density

1.00 g/mL at 25 °C

SMILES string

O=[Fe].O=[Fe]O[Fe]=O

InChI

1S/3Fe.4O

InChI key

SZVJSHCCFOBDDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Concentration 5mg/ml includes total weight nanocrystals plus ligands.
Iron oxide(II,III), magnetic nanoparticles solution is a chemically stable dispersed solution with a large surface to volume ratio. Its properties include good biocompatibility, and magnetic susceptibility (χ).

Application

Fe3O4 magnetic nanoparticles solution can be used in a variety of biomedical applications such as drug delivery. It can also be coated for immobilization of surface atoms for chelation of metal ions.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnetic nanoparticles-a review
Indira TK and Lakshmi PK
International Journal of Pharmaceutical Sciences and Nanotechnology, 3(3), 1035-1042 (2010)
Microfluidic synthesis of microfibers for magnetic-responsive controlled drug release and cell culture
Lin Y, et al.
Testing, 7(3), e33184-e33184 (2012)
Magnetic resonance imaging of ultrasmall superparamagnetic iron oxide-labeled exosomes from stem cells: a new method to obtain labeled exosomes
Busato A, et al.
International journal of nanomedicine, 11(3), 2481-2481 (2016)
Magnetic gamma-Fe2O3 nanoparticles coated with poly-l-cysteine for chelation of As (III), Cu (II), Cd (II), Ni (II), Pb (II) and Zn (II)
White BR, et al.
Journal of Hazardous Materials, 161(2-3), 848-853 (2009)
Maoquan Chu et al.
Biomaterials, 34(16), 4078-4088 (2013-03-08)
The photothermal effect of Fe3O4 magnetic nanoparticles is investigated for cancer therapy both in vitro and in vivo experiments. Heat is found to be rapidly generated by red and near-infrared (NIR) range laser irradiation of Fe3O4 nanoparticles with spherical, hexagonal and wire-like

Articles

Explore DNA-functionalized nanoparticles in sensors for precise target analyte detection. Learn about types, synthesis, functionalization, and design optimization, with insights into challenges and prospects.

Explore DNA-functionalized nanoparticles in sensors for precise target analyte detection. Learn about types, synthesis, functionalization, and design optimization, with insights into challenges and prospects.

Explore DNA-functionalized nanoparticles in sensors for precise target analyte detection. Learn about types, synthesis, functionalization, and design optimization, with insights into challenges and prospects.

Explore DNA-functionalized nanoparticles in sensors for precise target analyte detection. Learn about types, synthesis, functionalization, and design optimization, with insights into challenges and prospects.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service