Skip to Content
Merck
All Photos(1)

Key Documents

17938

Sigma-Aldrich

25-Hydroxyvitamin D3 monohydrate

≥99.0% (HPLC)

Synonym(s):

25-Hydroxycholecalciferol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O2 · H2O
CAS Number:
Molecular Weight:
418.65
Beilstein:
4270041
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058003
PubChem Substance ID:
NACRES:
NA.79
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Assay

≥99.0% (HPLC)

form

powder or crystals

color

white to off-white

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]1(CC[C@@]2([H])C(\CCC[C@]12C)=C\C=C3\C[C@@H](O)CCC3=C)[C@H](C)CCCC(C)(C)O

InChI

1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23+,24-,25+,27-/m1/s1

InChI key

JWUBBDSIWDLEOM-DTOXIADCSA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cholecalciferol is an inactive form of vitamin D3 which undergoes various levels of hydroxylation to form active vitamin D3 analogs. 1α-Hydroxyvitamin D3 (alfacalcidol) is a synthetic analog that is metabolized to 1,25-dihydroxycholecalciferol, the biologically active form of vitamin D3. Other analogues of cholecalciferol result from different hydroxylations. 25-dihydroxycholecalciferol is a the major circulating form of vitamin D3 and a physiological precursor of 1α,25-Dihydroxycholecalciferol. The conversion is catalyzed by 25-Hydroxyvitamin D3 1-α-hydroxylase (VD3 1A hydroxylase) also known ascytochrome p450 27B1 (CYP27B1).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew J Rowling et al.
The Journal of nutrition, 136(11), 2754-2759 (2006-10-24)
The major circulating form of vitamin D is 25-hydroxycholecalciferol [25(OH)D3], which is delivered to target tissues in complex with the serum vitamin D binding protein (DBP). We recently observed that mammary cells can metabolize 25(OH)D3 to 1,25-dihydroxycholecalciferol [1,25(OH)(2)D3], the vitamin
H F DeLuca
Biochemical Society transactions, 10(3), 147-158 (1982-06-01)
Cholecalciferol must be regarded as a pro-hormone rather than a vitamin, since it is normally produced in skin under the influence of ultraviolet light. Cholecalciferol must be metabolized in liver to 25-hydroxycholecalciferol and subsequently to 1,25-dihydroxycholecalciferol before it can act
Carly M Kemmis et al.
The Journal of nutrition, 136(4), 887-892 (2006-03-22)
1alpha,25-dihydroxycholecalciferol [1alpha,25(OH)2D3], the active form of cholecalciferol, is a negative growth regulator of breast cancer cells. CYP27B1 is a cytochrome P450-containing hydroxylase expressed in kidney and other tissues that generates 1alpha,25(OH)2D3 from an inactive vitamin D precursor 25-hydroxycholecalciferol [25(OH)D3]. In
Fadia S AlBuhairan et al.
The Journal of adolescent health : official publication of the Society for Adolescent Medicine, 57(3), 263-269 (2015-08-25)
With the increasing burden of noncommunicable disease, adolescence is viewed as an opportune time to prevent the onset of certain behaviors and promote healthy states. Although adolescents comprise a considerable portion of Saudi Arabia's population, they have received insufficient attention
Marina V Antunes et al.
Therapeutic drug monitoring, 37(6), 733-744 (2015-04-09)
In view of the large variability on therapeutic response and the multiple factors associated to tamoxifen (TAM) metabolic activation, this study aimed to evaluate the effect of CYP2D6 and CYP3A4 phenotypes, drug interactions, and vitamin D exposure on TAM metabolism

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service