Skip to Content
Merck
All Photos(3)

Key Documents

733490

Sigma-Aldrich

Antimony(III) telluride

greener alternative

powder, −325 mesh, 99.96% trace metals basis

Synonym(s):

Antimony sesquitelluride, Antimony telluride, Diantimony tritelluride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sb2Te3
CAS Number:
Molecular Weight:
626.32
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.96% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

−325 mesh

mp

629 °C

density

6.5 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Te]=[Sb][Te][Sb]=[Te]

InChI

1S/2Sb.3Te

InChI key

BPDQXJZWVBPDSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Antimony(III) telluride (Sb2Te3) is a three dimensional topological insulator that can be used as a binary sesquichalogenide. It forms p-type semiconducting films for the formation of thermoelectric materials.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Sb2Te3 in combination with bismuth(III) telluride (Bi2Te3) can form super-lattices, which facilitate the fabrication of devices such as thermoelectric generators.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pradyumnan, P. P.; Swathikrishnan
Indian Journal of Pure and Applied Physics, 48, 115-115 (2010)
Ambipolar field effect in the ternary topological insulator (Bi x Sb 1-x) 2 Te 3 by composition tuning
Kong D, et al.
Nature Nanotechnology, 6(11), 705-705 (2011)
Scherrer, H.; Scherrer, S.
CRC Handbook of Thermoelectronics, 211-211 (1995)
Observation of time-reversal-protected single-Dirac-cone topological-insulator states in Bi 2 Te 3 and Sb 2 Te 3
Hsieh D, et al.
Physical Review Letters, 103(14), 146401-146401 (2009)
A wearable thermoelectric generator fabricated on a glass fabric
Kim SJ, et al.
Energy & Environmental Science, 7(6), 1959-1965 (2014)

Articles

Thermoelectric materials comprise a wide range of solid compounds distinguished by their ability to convert thermal and electrical energy.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service